Thermo Scientific™
TSX Series High-Performance Plasma Freezers
Catalog number: TSX5030LA
Have Questions?
Thermo Scientific™
TSX Series High-Performance Plasma Freezers
Catalog number: TSX5030LA
Have Questions?
Thermo Scientific™ TSX Series high-performance –30°C plasma freezers are designed to meet strict safety and performance requirements established by the AABB and ANRC. 
Now certified to the AABB Standards-Compliant Product Evaluation (SCoPE) program, the FDA Class II and MDD 93/42/EEC Class IIa listed TSX series with V-Drive technology delivers the performance and quality you need in addition to the low sound, low energy and exceptional user experience you deserve. 
Catalog Number
TSX5030LA
Unit Size
Each
Plug Type
NEMA 5-15
Electrical Requirements
115V, 60Hz
Chart Recorder
Standard
Capacity (English)
51.1 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleDouble Solid
No. of Doors2
DrawersStainless Steel
No. of Drawers14
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.4 cm
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior56.5 in.
Width (English) Interior52.5 in.
Width (Metric) Exterior143.5 cm
Width (Metric) Interior133.4 cm
Monitoring OptionsWireless
Chart RecorderStandard
Smart View CompatibilitySmart-Vue Compatible
Amperage20 A (CB)
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
HoldsBoxes (1064)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 5-15
Electrical Requirements
115V, 60Hz
Chart Recorder
Standard
Capacity (English)
23 cu. ft.
Price (USD)
Full specifications
TypePlasma Freezer
Capacity (English)23 cu. ft.
Capacity (Metric)650 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
Dimensions (L x W x H) Exterior37.9 x 28 x 78.6 in. (96.2 x 71.1 x 199.6 cm)
Depth (English) Exterior37.9 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.2 cm
Depth (Metric) Interior72.3 cm
Height (English) Exterior78.6 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.6 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior28 in.
Width (English) Interior24 in.
Width (Metric) Exterior71.1 cm
Width (Metric) Interior61.1 cm
Monitoring OptionsWireless
Chart RecorderStandard
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)526 lb.
Shipping Weight (Metric)239 kg
Weight (English)461 lb.
Weight (Metric)209 kg
HoldsBoxes (532)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
CEE 7/7
Electrical Requirements
208-230V, 50Hz
Chart Recorder
Standard
Capacity (English)
51.1 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleDouble Solid
No. of Doors2
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, cUL, CE, FDA, MDD 93/42/EEC (Class IIa), AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior37.9 x 56.5 x 78.5 in. (96.2 x 143.5 x 199.4 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.4 cm
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior56.5 in.
Width (English) Interior52.5 in.
Width (Metric) Exterior143.5 cm
Width (Metric) Interior133.4 cm
Monitoring OptionsWireless
Chart RecorderStandard
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A (CB)
Plug TypeCEE 7/7
Voltage230 V
Electrical Requirements208-230V, 50Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
HoldsBoxes (1064)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 6-15
Electrical Requirements
208-230V, 60Hz
Chart Recorder
Optional
Capacity (English)
29.2 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.3 cm
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior34.0 in.
Width (English) Interior30 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior76.2 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208-230V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)556 lb.
Shipping Weight (Metric)252 kg
Weight (English)488 lb.
Weight (Metric)221 kg
HoldsBoxes (665)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 6-15
Electrical Requirements
208-230V, 60Hz
Chart Recorder
Optional
Capacity (English)
11.5 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Depth (English) Exterior31.1 in.
Depth (English) Interior21.8 in.
Depth (Metric) Exterior79.0 cm
Depth (Metric) Interior55.4 cm
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Exterior24.0 in.
Width (English) Interior20 in.
Width (Metric) Exterior61.9 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208-230V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)412 lb.
Shipping Weight (Metric)187 kg
Weight (English)358 lb.
Weight (Metric)162 kg
HoldsBoxes (273)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 5-15
Electrical Requirements
115V, 60Hz
Chart Recorder
Optional
Capacity (English)
11.5 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Depth (English) Exterior31.1 in.
Depth (English) Interior21.8 in.
Depth (Metric) Exterior79.0 cm
Depth (Metric) Interior55.4 cm
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Exterior24.0 in.
Width (English) Interior20 in.
Width (Metric) Exterior61.9 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)412 lb.
Shipping Weight (Metric)187 kg
Weight (English)358 lb.
Weight (Metric)162 kg
HoldsBoxes (273)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 5-15
Electrical Requirements
115V, 60Hz
Chart Recorder
Optional
Capacity (English)
29.2 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.3 cm
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior34.0 in.
Width (English) Interior30 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior76.2 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)556 lb.
Shipping Weight (Metric)252 kg
Weight (English)488 lb.
Weight (Metric)221 kg
HoldsBoxes (665)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
CEE 7/7
Electrical Requirements
208-230V, 50Hz
Chart Recorder
Optional
Capacity (English)
11.5 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, cUL, CE, FDA, MDD 93/42/EEC (Class IIa), AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Depth (English) Exterior31.1 in.
Depth (English) Interior21.8 in.
Depth (Metric) Exterior79.0 cm
Depth (Metric) Interior55.4 cm
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Exterior24.0 in.
Width (English) Interior20 in.
Width (Metric) Exterior61.9 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeCEE 7/7
Voltage230 V
Electrical Requirements208-230V, 50Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)412 lb.
Shipping Weight (Metric)187 kg
Weight (English)358 lb.
Weight (Metric)162 kg
HoldsBoxes (273)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
NEMA 6-15
Electrical Requirements
208-230V, 60Hz
Chart Recorder
Standard
Capacity (English)
51.1 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleDouble Solid
No. of Doors2
DrawersStainless Steel
No. of Drawers14
Certifications/ComplianceUL, cUL, ENERGYSTAR, FDA
DisplayMicroprocessor
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.4 cm
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior56.5 in.
Width (English) Interior52.5 in.
Width (Metric) Exterior143.5 cm
Width (Metric) Interior133.4 cm
Monitoring OptionsWireless
Chart RecorderStandard
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A (CB)
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208-230V, 60Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
HoldsBoxes (1064)
Rack Capacity7
Unit SizeEach
Catalog Number
Unit Size
Each
Plug Type
CEE 7/7
Electrical Requirements
208-230V, 50Hz
Chart Recorder
Optional
Capacity (English)
29.2 cu. ft.
Price (USD)
Full specifications
TypeHigh-Performance Freezer
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, cUL, CE, FDA, MDD 93/42/EEC (Class IIa), AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.3 cm
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior34.0 in.
Width (English) Interior30 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior76.2 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeCEE 7/7
Voltage230 V
Electrical Requirements208-230V, 50Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)556 lb.
Shipping Weight (Metric)252 kg
Weight (English)488 lb.
Weight (Metric)221 kg
HoldsBoxes (665)
Rack Capacity7
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizePlug TypeElectrical RequirementsChart RecorderCapacity (English)Price (USD)
TSX5030LAFull specifications
EachNEMA 5-15115V, 60HzStandard51.1 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleDouble Solid
No. of Doors2
DrawersStainless Steel
No. of Drawers14
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.4 cm
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior56.5 in.
Width (English) Interior52.5 in.
Width (Metric) Exterior143.5 cm
Width (Metric) Interior133.4 cm
Monitoring OptionsWireless
Chart RecorderStandard
Smart View CompatibilitySmart-Vue Compatible
Amperage20 A (CB)
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
HoldsBoxes (1064)
Rack Capacity7
Unit SizeEach
TSX2330LAFull specifications
EachNEMA 5-15115V, 60HzStandard23 cu. ft.Request A Quote
TypePlasma Freezer
Capacity (English)23 cu. ft.
Capacity (Metric)650 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior28.5 x 24 x 58 in. (72.3 x 61 x 147.3 cm)
Dimensions (L x W x H) Exterior37.9 x 28 x 78.6 in. (96.2 x 71.1 x 199.6 cm)
Depth (English) Exterior37.9 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.2 cm
Depth (Metric) Interior72.3 cm
Height (English) Exterior78.6 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.6 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior28 in.
Width (English) Interior24 in.
Width (Metric) Exterior71.1 cm
Width (Metric) Interior61.1 cm
Monitoring OptionsWireless
Chart RecorderStandard
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)526 lb.
Shipping Weight (Metric)239 kg
Weight (English)461 lb.
Weight (Metric)209 kg
HoldsBoxes (532)
Rack Capacity7
Unit SizeEach
TSX5030LVFull specifications
EachCEE 7/7208-230V, 50HzStandard51.1 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleDouble Solid
No. of Doors2
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, cUL, CE, FDA, MDD 93/42/EEC (Class IIa), AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior37.9 x 56.5 x 78.5 in. (96.2 x 143.5 x 199.4 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.4 cm
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior56.5 in.
Width (English) Interior52.5 in.
Width (Metric) Exterior143.5 cm
Width (Metric) Interior133.4 cm
Monitoring OptionsWireless
Chart RecorderStandard
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A (CB)
Plug TypeCEE 7/7
Voltage230 V
Electrical Requirements208-230V, 50Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
HoldsBoxes (1064)
Rack Capacity7
Unit SizeEach
TSX3030LDFull specifications
EachNEMA 6-15208-230V, 60HzOptional29.2 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.3 cm
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior34.0 in.
Width (English) Interior30 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior76.2 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208-230V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)556 lb.
Shipping Weight (Metric)252 kg
Weight (English)488 lb.
Weight (Metric)221 kg
HoldsBoxes (665)
Rack Capacity7
Unit SizeEach
TSX1230LDFull specifications
EachNEMA 6-15208-230V, 60HzOptional11.5 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Depth (English) Exterior31.1 in.
Depth (English) Interior21.8 in.
Depth (Metric) Exterior79.0 cm
Depth (Metric) Interior55.4 cm
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Exterior24.0 in.
Width (English) Interior20 in.
Width (Metric) Exterior61.9 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208-230V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)412 lb.
Shipping Weight (Metric)187 kg
Weight (English)358 lb.
Weight (Metric)162 kg
HoldsBoxes (273)
Rack Capacity7
Unit SizeEach
TSX1230LAFull specifications
EachNEMA 5-15115V, 60HzOptional11.5 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Depth (English) Exterior31.1 in.
Depth (English) Interior21.8 in.
Depth (Metric) Exterior79.0 cm
Depth (Metric) Interior55.4 cm
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Exterior24.0 in.
Width (English) Interior20 in.
Width (Metric) Exterior61.9 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)412 lb.
Shipping Weight (Metric)187 kg
Weight (English)358 lb.
Weight (Metric)162 kg
HoldsBoxes (273)
Rack Capacity7
Unit SizeEach
TSX3030LAFull specifications
EachNEMA 5-15115V, 60HzOptional29.2 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, ENERGYSTAR, cUL, FDA, AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.3 cm
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior34.0 in.
Width (English) Interior30 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior76.2 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115V, 60Hz
Frequency60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)556 lb.
Shipping Weight (Metric)252 kg
Weight (English)488 lb.
Weight (Metric)221 kg
HoldsBoxes (665)
Rack Capacity7
Unit SizeEach
TSX1230LVFull specifications
EachCEE 7/7208-230V, 50HzOptional11.5 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)11.5 cu. ft.
Capacity (Metric)326 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, cUL, CE, FDA, MDD 93/42/EEC (Class IIa), AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Depth (English) Exterior31.1 in.
Depth (English) Interior21.8 in.
Depth (Metric) Exterior79.0 cm
Depth (Metric) Interior55.4 cm
Height (English) Exterior73.0 in.
Height (English) Interior52.4 in.
Height (Metric) Exterior185.4 cm
Height (Metric) Interior133.1 cm
Width (English) Exterior24.0 in.
Width (English) Interior20 in.
Width (Metric) Exterior61.9 cm
Width (Metric) Interior50.8 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeCEE 7/7
Voltage230 V
Electrical Requirements208-230V, 50Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)412 lb.
Shipping Weight (Metric)187 kg
Weight (English)358 lb.
Weight (Metric)162 kg
HoldsBoxes (273)
Rack Capacity7
Unit SizeEach
TSX5030LDFull specifications
EachNEMA 6-15208-230V, 60HzStandard51.1 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleDouble Solid
No. of Doors2
DrawersStainless Steel
No. of Drawers14
Certifications/ComplianceUL, cUL, ENERGYSTAR, FDA
DisplayMicroprocessor
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostAutomatic
Dimensions (D x W x H) Interior28.5 x 52.5 x 58 in. (72.4 x 133.4 x 147.3 cm)
Dimensions (L x W x H) Exterior37.8 x 56.5 x 78.5 in. (96 x 143.5 x 199.4 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.4 cm
Height (English) Exterior78.5 in.
Height (English) Interior58 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior56.5 in.
Width (English) Interior52.5 in.
Width (Metric) Exterior143.5 cm
Width (Metric) Interior133.4 cm
Monitoring OptionsWireless
Chart RecorderStandard
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A (CB)
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208-230V, 60Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
HoldsBoxes (1064)
Rack Capacity7
Unit SizeEach
TSX3030LVFull specifications
EachCEE 7/7208-230V, 50HzOptional29.2 cu. ft.Request A Quote
TypeHigh-Performance Freezer
Capacity (English)29.2 cu. ft.
Capacity (Metric)827 L
Temperature (Setpoint)-35° to -15°C (-30°C)
Door StyleSolid
No. of Doors1
DrawersStainless Steel
No. of Drawers7
Certifications/ComplianceUL, cUL, CE, FDA, MDD 93/42/EEC (Class IIa), AABB SCoPE
DisplayMicroprocessor
Product LineTSX Series
DefrostAutomatic
Dimensions (DxWxH) Exterior37.9 x 34.0 x 78.5 in. (96.2 x 86.4 x 199.4 cm)
Dimensions (D x W x H) Interior28.5 x 30.0 x 58.0 in. ( 72.3 x 76.2 x 147.3 cm)
Depth (English) Exterior37.8 in.
Depth (English) Interior28.5 in.
Depth (Metric) Exterior96.0 cm
Depth (Metric) Interior72.3 cm
Height (English) Exterior78.5 in.
Height (English) Interior58.0 in.
Height (Metric) Exterior199.4 cm
Height (Metric) Interior147.3 cm
Width (English) Exterior34.0 in.
Width (English) Interior30 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior76.2 cm
Monitoring OptionsWireless
Chart RecorderOptional
Smart View CompatibilitySmart-Vue Compatible
Amperage15 A
Plug TypeCEE 7/7
Voltage230 V
Electrical Requirements208-230V, 50Hz
Frequency50/60 Hz
Green FeaturesEnergy efficient
RefrigerantR290
Shipping Weight (English)556 lb.
Shipping Weight (Metric)252 kg
Weight (English)488 lb.
Weight (Metric)221 kg
HoldsBoxes (665)
Rack Capacity7
Unit SizeEach
Showing 10 of 12Show more
The drive for sample integrity and security
  • The unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance
Enabling compliance
  • Certified to AABB SCoPE
  • Listed as US FDA Class II and/or MDD 93/42/EEC Class IIa* Medical Devices
  • Tested and certified to conform with the design and safety standards of UL/EN 61010, and CE Directive 2011/65/EU and 2015/863
Designed for an ergonomic and comfortable experience
  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Convenient, fully extendable and adjustable stainless-steel drawers
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Self-closing door with 90° stop to assist with inventory loads
  • Standard, built-in chart recorder
Verified sustainability
  • US EPA ENERGY STAR Certified with up to 40% less energy usage than conventional refrigerant models
  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, US Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility
Ready for connectivity and monitoring
  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.**
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
Make it your own
  • Alternate Door swing configuration and door types
  • Stainless Steel interior finish
  • E-Lock Adapter kits 
  • 4-20 milliamp transmitters and other remote alarm modules
  • Specialty, regional, and country specific plugs
Services and support
  • Industry Leading 120-month compressor warranty (varies by country, please contact us for details)
  • Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs
*V voltage models only
**DeviceLink Connect availability is country specific. Please contact your Thermo Scientific representative for more details.