Search Thermo Fisher Scientific
Catalog Number | Door Style | Electrical Requirements | Plug Type |
---|---|---|---|
TSX505SA | Single Solid | 100 - 240 V 50-60 Hz | NEMA 5-15 P, More Global Types Available |
TSX505GA | Single Glass | 100 - 240 V 50-60 Hz | NEMA 5-15 P, More Global Types Available |
TSX505SAVK | Single Solid | 100 - 240 V 50-60 Hz | NEMA 5-15 P |
Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2° to 8°C. The Series' V-drive is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Unique to the TSX505, our dual refrigeration solution operates through the action of two synchronized solid-state heat pumps. The two solid-state heat pumps work to cool a combination of cold wall and forced air to help ensure that conditions are optimal for the most sensitive applications
Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.