Informational:Informational:Our newest TSX Universal Ultra-Low Temperature Freezers have launched. Visit the following page to learn more: TSX70086FA
Thermo Scientific™
TSX Series Ultra-Low Freezers
Catalog number: TSX60086A
Have Questions?
Thermo Scientific™
TSX Series Ultra-Low Freezers
Catalog number: TSX60086A
Have Questions?
The ENERGY STAR™ certified Thermo Scientific™ TSX Series ultra-low freezers are designed to meet the highest standards in reliability, sustainability and temperature management. At the heart of the TSX Series is V-drive technology, designed to adapt to user patterns, substantially reducing energy and HVAC costs without compromising performance. All TSX Series freezers are manufactured in a zero-waste facility and are built with natural refrigerants and water-blown foam insulation, compliant with the U.S. EPA’s Significant New Alternatives Policy (SNAP), the European Union’s F-Gas compliance and other sustainability standards.
From our first ULT freezer in 1940 to today, the combined experience of REVCO, Forma Scientific, Jewett, Harris, and other families of products continues to drive our ability to deliver industry-leading innovations that help accelerate science and health care.
Catalog Number
TSX60086A
Unit Size
Each
Holds
2 in. Boxes (600)
Dimensions (DxWxH) Exterior
37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Electrical Requirements
115 V 60 Hz
Rack Capacity
24
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 600-Box Capacity, 115V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)28.8 cu. ft.
Capacity (Metric)816 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf245 lb.
Capacity (Metric) Shelf111 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 34.4 x 51.2 in. (71.9 x 87.3 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior39.6 in.
Width (English) Interior34.4 in.
Width (Metric) Exterior100.6 cm
Width (Metric) Interior87.3 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage8.5/15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity60,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)790 lb.
Shipping Weight (Metric)358 kg
Weight (English)679 lb.
Weight (Metric)308 kg
Holds2 in. Boxes (600)
Rack Capacity24
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (400)
Dimensions (DxWxH) Exterior
37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Electrical Requirements
115 V 60 Hz
Rack Capacity
16
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 400-Box Capacity, 115V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)19.4 cu. ft.
Capacity (Metric)549 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf165 lb.
Capacity (Metric) Shelf75 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 23.1 x 51.2 in. (71.9 x 58.8 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior28.3 in.
Width (English) Interior23.1 in.
Width (Metric) Exterior71.9 cm
Width (Metric) Interior58.8 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage9.0/15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy Efficient
Vial Capacity40,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)665 lb.
Shipping Weight (Metric)302 kg
Weight (English)573 lb.
Weight (Metric)260 kg
Holds2 in. Boxes (400)
Rack Capacity16
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (400)
Dimensions (DxWxH) Exterior
37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Electrical Requirements
208 - 230 V 60 Hz
Rack Capacity
16
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 400-Box Capacity, 208-230V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)19.4 cu. ft.
Capacity (Metric)549 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf165 lb.
Capacity (Metric) Shelf75 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 23.1 x 51.2 in.(71.9 x 58.8 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior28.3 in.
Width (English) Interior23.1 in.
Width (Metric) Exterior71.9 cm
Width (Metric) Interior58.8 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage4/10 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity40,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)640 lb.
Shipping Weight (Metric)290 kg
Weight (English)573 lb.
Weight (Metric)260 kg
Holds2 in. Boxes (400)
Rack Capacity16
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (400)
Dimensions (DxWxH) Exterior
37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Electrical Requirements
230 V 50 Hz
Rack Capacity
16
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 400-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)19.4 cu. ft.
Capacity (Metric)549 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf165 lb.
Capacity (Metric) Shelf75 kg
Certifications/ComplianceCE, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 23.1 x 51.2 in. (71.9 x 58.8 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior28.3 in.
Width (English) Interior23.1 in.
Width (Metric) Exterior71.9 cm
Width (Metric) Interior58.8 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage4/10 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Green FeaturesEnergy efficient
Vial Capacity40,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)640 lb.
Shipping Weight (Metric)290 kg
Weight (English)573 lb.
Weight (Metric)260 kg
Holds2 in. Boxes (400)
Rack Capacity16
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (500)
Dimensions (DxWxH) Exterior
37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Electrical Requirements
115 V 60 Hz
Rack Capacity
20
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 500-Box Capacity, 115V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)24.1 cu. ft.
Capacity (Metric)682 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf205 lb.
Capacity (Metric) Shelf93 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 28.8 x 51.2 in. (71.9 x 73.0 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior34 in.
Width (English) Interior28.8 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior73.0 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage9.5/15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity50,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)721 lb.
Shipping Weight (Metric)327 kg
Weight (English)627 lb.
Weight (Metric)284 kg
Holds2 in. Boxes (500)
Rack Capacity20
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (500)
Dimensions (DxWxH) Exterior
37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Electrical Requirements
208 - 230 V 60 Hz
Rack Capacity
20
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 500-Box Capacity, 208-230V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)24.1 cu. ft.
Capacity (Metric)682 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf205 lb.
Capacity (Metric) Shelf93 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 28.8 x 51.2 in. (71.9 x 73.0 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior34 in.
Width (English) Interior28.8 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior73.0 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage5.1/10 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity50,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)696 lb.
Shipping Weight (Metric)316 kg
Weight (English)627 lb.
Weight (Metric)284 kg
Holds2 in. Boxes (500)
Rack Capacity20
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (500)
Dimensions (DxWxH) Exterior
37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Electrical Requirements
230 V 50 Hz
Rack Capacity
20
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 500-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)24.1 cu. ft.
Capacity (Metric)682 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf205 lb.
Capacity (Metric) Shelf93 kg
Certifications/ComplianceCE, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 28.8 x 51.2 in. (71.9 x 73.0 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior34 in.
Width (English) Interior28.8 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior73.0 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage5.2/10 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Green FeaturesEnergy efficient
Vial Capacity50,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)696 lb.
Shipping Weight (Metric)316 kg
Weight (English)627 lb.
Weight (Metric)284 kg
Holds2 in. Boxes (500)
Rack Capacity20
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (600)
Dimensions (DxWxH) Exterior
37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Electrical Requirements
208 - 230 V 60 Hz
Rack Capacity
24
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 600-Box Capacity, 208-230V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)28.8 cu. ft.
Capacity (Metric)816 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf245 lb.
Capacity (Metric) Shelf111 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 34.4 x 51.2 in.(71.9 x 87.3 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior39.6 in.
Width (English) Interior34.4 in.
Width (Metric) Exterior100.6 cm
Width (Metric) Interior87.3 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage4/10 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity60,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)765 lb.
Shipping Weight (Metric)347 kg
Weight (English)679 lb.
Weight (Metric)308 kg
Holds2 in. Boxes (600)
Rack Capacity24
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (600)
Dimensions (DxWxH) Exterior
37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Electrical Requirements
230 V 50 Hz
Rack Capacity
24
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 600-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)28.8 cu. ft.
Capacity (Metric)816 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf245 lb.
Capacity (Metric) Shelf111 kg
Certifications/ComplianceCE, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 34.4 x 51.2 in. (71.9 x 87.3 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior39.6 in.
Width (English) Interior34.4 in.
Width (Metric) Exterior100.6 cm
Width (Metric) Interior87.3 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage4/10 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Green FeaturesEnergy efficient
Vial Capacity60,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)765 lb.
Shipping Weight (Metric)347 kg
Weight (English)679 lb.
Weight (Metric)308 kg
Holds2 in. Boxes (600)
Rack Capacity24
Unit SizeEach
Catalog Number
Unit Size
Each
Holds
2 in. Boxes (700)
Dimensions (DxWxH) Exterior
37.6 x 45.2 x 78 in. (95.5 x 114.8 x 198.1 cm)
Electrical Requirements
115 V 60 Hz
Rack Capacity
28
Price (USD)
Full specifications
DescriptionTSX Series Ultra-Low Freezers, 700-Box Capacity, 115V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)33.5 cu. ft.
Capacity (Metric)949 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf285 lb.
Capacity (Metric) Shelf129 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 45.2 x 78 in. (95.5 x 114.8 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 40.1 x 51.2 in. (71.9 x 101.9 x 130.0cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior45.2 in.
Width (English) Interior40 in.
Width (Metric) Exterior114.8 cm
Width (Metric) Interior101.6 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage10.6/15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity70,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)894 lb.
Shipping Weight (Metric)406 kg
Weight (English)778 lb.
Weight (Metric)353 kg
Holds2 in. Boxes (700)
Rack Capacity28
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeHoldsDimensions (DxWxH) ExteriorElectrical RequirementsRack CapacityPrice (USD)
TSX60086AFull specifications
Each2 in. Boxes (600)37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)115 V 60 Hz24Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 600-Box Capacity, 115V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)28.8 cu. ft.
Capacity (Metric)816 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf245 lb.
Capacity (Metric) Shelf111 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 34.4 x 51.2 in. (71.9 x 87.3 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior39.6 in.
Width (English) Interior34.4 in.
Width (Metric) Exterior100.6 cm
Width (Metric) Interior87.3 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage8.5/15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity60,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)790 lb.
Shipping Weight (Metric)358 kg
Weight (English)679 lb.
Weight (Metric)308 kg
Holds2 in. Boxes (600)
Rack Capacity24
Unit SizeEach
TSX40086AFull specifications
Each2 in. Boxes (400)37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)115 V 60 Hz16Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 400-Box Capacity, 115V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)19.4 cu. ft.
Capacity (Metric)549 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf165 lb.
Capacity (Metric) Shelf75 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 23.1 x 51.2 in. (71.9 x 58.8 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior28.3 in.
Width (English) Interior23.1 in.
Width (Metric) Exterior71.9 cm
Width (Metric) Interior58.8 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage9.0/15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy Efficient
Vial Capacity40,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)665 lb.
Shipping Weight (Metric)302 kg
Weight (English)573 lb.
Weight (Metric)260 kg
Holds2 in. Boxes (400)
Rack Capacity16
Unit SizeEach
TSX40086DFull specifications
Each2 in. Boxes (400)37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)208 - 230 V 60 Hz16Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 400-Box Capacity, 208-230V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)19.4 cu. ft.
Capacity (Metric)549 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf165 lb.
Capacity (Metric) Shelf75 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 23.1 x 51.2 in.(71.9 x 58.8 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior28.3 in.
Width (English) Interior23.1 in.
Width (Metric) Exterior71.9 cm
Width (Metric) Interior58.8 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage4/10 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity40,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)640 lb.
Shipping Weight (Metric)290 kg
Weight (English)573 lb.
Weight (Metric)260 kg
Holds2 in. Boxes (400)
Rack Capacity16
Unit SizeEach
TSX40086VFull specifications
Each2 in. Boxes (400)37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)230 V 50 Hz16Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 400-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)19.4 cu. ft.
Capacity (Metric)549 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf165 lb.
Capacity (Metric) Shelf75 kg
Certifications/ComplianceCE, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 28.3 x 78 in. (95.5 x 71.9 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 23.1 x 51.2 in. (71.9 x 58.8 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior28.3 in.
Width (English) Interior23.1 in.
Width (Metric) Exterior71.9 cm
Width (Metric) Interior58.8 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage4/10 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Green FeaturesEnergy efficient
Vial Capacity40,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)640 lb.
Shipping Weight (Metric)290 kg
Weight (English)573 lb.
Weight (Metric)260 kg
Holds2 in. Boxes (400)
Rack Capacity16
Unit SizeEach
TSX50086AFull specifications
Each2 in. Boxes (500)37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)115 V 60 Hz20Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 500-Box Capacity, 115V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)24.1 cu. ft.
Capacity (Metric)682 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf205 lb.
Capacity (Metric) Shelf93 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 28.8 x 51.2 in. (71.9 x 73.0 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior34 in.
Width (English) Interior28.8 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior73.0 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage9.5/15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity50,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)721 lb.
Shipping Weight (Metric)327 kg
Weight (English)627 lb.
Weight (Metric)284 kg
Holds2 in. Boxes (500)
Rack Capacity20
Unit SizeEach
TSX50086DFull specifications
Each2 in. Boxes (500)37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)208 - 230 V 60 Hz20Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 500-Box Capacity, 208-230V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)24.1 cu. ft.
Capacity (Metric)682 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf205 lb.
Capacity (Metric) Shelf93 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 28.8 x 51.2 in. (71.9 x 73.0 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior34 in.
Width (English) Interior28.8 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior73.0 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage5.1/10 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity50,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)696 lb.
Shipping Weight (Metric)316 kg
Weight (English)627 lb.
Weight (Metric)284 kg
Holds2 in. Boxes (500)
Rack Capacity20
Unit SizeEach
TSX50086VFull specifications
Each2 in. Boxes (500)37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)230 V 50 Hz20Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 500-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)24.1 cu. ft.
Capacity (Metric)682 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf205 lb.
Capacity (Metric) Shelf93 kg
Certifications/ComplianceCE, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 34.0 x 78 in. (95.5 x 86.4 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 28.8 x 51.2 in. (71.9 x 73.0 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior34 in.
Width (English) Interior28.8 in.
Width (Metric) Exterior86.4 cm
Width (Metric) Interior73.0 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage5.2/10 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Green FeaturesEnergy efficient
Vial Capacity50,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)696 lb.
Shipping Weight (Metric)316 kg
Weight (English)627 lb.
Weight (Metric)284 kg
Holds2 in. Boxes (500)
Rack Capacity20
Unit SizeEach
TSX60086DFull specifications
Each2 in. Boxes (600)37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)208 - 230 V 60 Hz24Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 600-Box Capacity, 208-230V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)28.8 cu. ft.
Capacity (Metric)816 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf245 lb.
Capacity (Metric) Shelf111 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 34.4 x 51.2 in.(71.9 x 87.3 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior39.6 in.
Width (English) Interior34.4 in.
Width (Metric) Exterior100.6 cm
Width (Metric) Interior87.3 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage4/10 A
Plug TypeNEMA 6-15
Voltage208/230 V
Electrical Requirements208 - 230 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity60,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)765 lb.
Shipping Weight (Metric)347 kg
Weight (English)679 lb.
Weight (Metric)308 kg
Holds2 in. Boxes (600)
Rack Capacity24
Unit SizeEach
TSX60086VFull specifications
Each2 in. Boxes (600)37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)230 V 50 Hz24Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 600-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)28.8 cu. ft.
Capacity (Metric)816 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf245 lb.
Capacity (Metric) Shelf111 kg
Certifications/ComplianceCE, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 34.4 x 51.2 in. (71.9 x 87.3 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior39.6 in.
Width (English) Interior34.4 in.
Width (Metric) Exterior100.6 cm
Width (Metric) Interior87.3 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage4/10 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Green FeaturesEnergy efficient
Vial Capacity60,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)765 lb.
Shipping Weight (Metric)347 kg
Weight (English)679 lb.
Weight (Metric)308 kg
Holds2 in. Boxes (600)
Rack Capacity24
Unit SizeEach
TSX70086AFull specifications
Each2 in. Boxes (700)37.6 x 45.2 x 78 in. (95.5 x 114.8 x 198.1 cm)115 V 60 Hz28Request A Quote
DescriptionTSX Series Ultra-Low Freezers, 700-Box Capacity, 115V, 60Hz
TypeUltra-Low Freezer, Upright
Capacity (English)33.5 cu. ft.
Capacity (Metric)949 L
Temperature Range-50°C to -86°C
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf285 lb.
Capacity (Metric) Shelf129 kg
Certifications/CompliancecULus, ENERGYSTAR
StyleUpright
DisplayTouch-Screen LCD
Product LineTSX Series
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior37.6 x 45.2 x 78 in. (95.5 x 114.8 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 40.1 x 51.2 in. (71.9 x 101.9 x 130.0cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Height (English) Exterior78 in.
Height (English) Interior51.2 in.
Height (Metric) Exterior198.1 cm
Height (Metric) Interior130.1 cm
Width (English) Exterior45.2 in.
Width (English) Interior40 in.
Width (Metric) Exterior114.8 cm
Width (Metric) Interior101.6 cm
Monitoring OptionsAlarm, Recorder
ConnectivityThermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP)
Smart View CompatibilitySmart-Vue Compatible
Amperage10.6/15 A
Plug TypeNEMA 5-15
Voltage115 V
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Green FeaturesEnergy efficient
Vial Capacity70,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)894 lb.
Shipping Weight (Metric)406 kg
Weight (English)778 lb.
Weight (Metric)353 kg
Holds2 in. Boxes (700)
Rack Capacity28
Unit SizeEach
Showing 10 of 12Show more
Performance
  • No need to worry about where your samples are stored within the freezer.  With peak variation less than 5°C, your samples are protected and safely stored throughout the entire cabinet space. *
  • Long warm-up time from -50°C to -80°C: 5.05 hours*
  • Quiet enough to reside directly in the laboratory, rather than in the hall. At just 45.5 dBA, the TSX Series’ sound level is comparable to a home refrigerator.*
  • V-drive’s adaptive control is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
Wireless Monitoring Solution
Thermo Scientific™ DeviceLink™ comes embedded in your ULT freezer  providing real-time monitoring of system status and sample temperature.
  • View critical lab equipment data and equipment notifications on your mobile and web applications.
    • Display temperature
    • Ambient temperature
    • Alarm and alerts notification
  • Access data anytime, anywhere—data and alarms from monitored lab equipment are securely stored on the Cloud and sent to the user via email and mobile-phone notifications
  • InstrumentConnect™ streamlined dashboard-style reporting reduces time spent interpreting data
  • Unlike traditional connectivity methods, DeviceLink™ operates on a client-cloud architecture, requiring minimal IT support
  • No need to install sensors—the only plug-in is the equipment data port to read the parameters directly from the factory-installed sensors
  • Applicable Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP
User Friendly
  • Newly re-designed touch-screen user interface with easy-to-use: set-up wizard, event log, temperature mapping and data downloads via a built-in USB port
  • Eliminate the need for a separate chart recorder with the touch-screen’s datalogging and temperature mapping features, all exportable via a built-in USB port
  • Ergonomic design including comfortable door handle, eye-level touch-screen and shelf-level placement that don’t require floor-level loading of racks
  • Four capacities (400, 500, 600 and 700 2-inch box) to accommodate a variety of storage needs and lab footprint specifications
  • Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes
  • Security features including alarm settings, padlock-compatible doors and optional access card entry
Sustainability
  • Completed the ACT Environmental Impact Factor Label labeling process, a third party–verified information about the environmental impact of laboratory products.
  • TSX ULT portfolio is ENERGY STAR™ certified providing significantly increased energy savings and decreased operational costs
  • Natural refrigerants and water-blown foam insulation, making the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), the European Union’s F-Gas and other sustainability standards
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Lower HVAC costs as a result of less heat emission into the environment
  • Upon receiving the unit, we advise you to properly dispose of any packaging material, used to secure the unit during the shipping process.  Please refer to local recycling codes and dispose of any materials accordingly.  For information on waste disposal sites, please refer to your local waste provider.
Footnotes:
* Typical freezer data based on internal testing with freezer setpoint at -80°C and ambient temperature at 20°C. Performance data varies across freezer size and electrical specifications.
Performance Data*:
  High Performance Mode Standard Mode    
 Model Energy ConsumptionPeak Variation from Setpoint Energy Consumption 2Peak Variation from Setpoint 2Sound1-min Door Opening Recovery to -75°CWarm Up Time (-80°C to -50°C)ENERGY STAR
UOM(kW-hr/day)(°C)(kW-hr/day)(°C)(dBa)(min)(min)(kW-hr/day cuft)
 TSX40086A 8.5 +8.2 / +0.2 10.7 +5.0 / -1.8 49.8 21 292 0.492
 TSX40086D 7.7 +7.5 / +0.5 9.9 +4.7 / -2.1 49.8 19 292 0.492
 TSX40086V 7.7 +7.5 / +0.5 9.9 +4.7 / -2.1 49.8 19 292 0.492
 TSX50086A 8.7 +8.0 / -0.1 11.4 +4.9 / -2.0 50.0 24 264 0.448
 TSX50086D 8.5 +8.6 / +0.2 10.8 +4.5 / -2.4 50.0 26 264 0.425
 TSX50086V 8.5 +8.6 / +0.2 10.8 +4.5 / -2.4 50.0 26 264 0.425
 TSX60086A 9.4 +8.0  /+1.9 11.5 +4.8 / -0.7 49.4 31 297 0.395
TSX60086D 8.6 +7.6 / +1.8 11.1 +4.5 / -0.3 49.4 31 297 0.336
TSX60086V 8.6 +7.6 / +1.8 11.1 +4.5 / -0.3 49.4 31 297 0.336
TSX70086A 10.2 +6.7 / +2.0 13.5 +4.8 / -1.0 48.6 29 339 0.359
TSX70086D 9.4 +7.4 / +2.0 12.7 +3.8 / -1.5 48.6 34 339 0.359
TSX70086V 9.4 +7.4 / +2.0 12.7 +3.8 / -1.5 48.6 34 339 0.359
  1. Data is representative of performance and not published product specifications. Results should not be referenced in developing any regulatory parameters for specific customer usage.
  2. Freezer performance will vary based on customer product volume, storage format, operating conditions, test methodology and performing recommended maintenance
  3. Continuous product enhancements may, without notice, result in amendments or omissions to this technical data sheet. Thermo Scientific cannot accept responsibility for damage, injury, loss or expenses resulting from misapplication of the information herein.