Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar).
Specifications
Certifications/ComplianceROHS
Length (English)0.15 in.
Length (Metric)0.38 cm
Max. Pressure20,000 psi
Port Style10 to 32 coned
Quantity10/Pk.
For Use With (Equipment)0.062 in. OD Tubing
MaterialStainless Steel
TypeMale Hex Nut
Unit SizePack of 10
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.