Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Actual product may vary
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Stainless Steel Unions, Tees and Crosses
Thermo Scientific™
Stainless Steel Unions, Tees and Crosses
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDescriptionSwept Volume
U-412Union0.020 in. through hole0.129 μL
U-402Union0.020 in. through hole0.134 μL
U-428Tee0.020 in. through hole0.57 μL
Catalog number U-412
Price (KRW)
-
Type:
Union
Description:
0.020 in. through hole
Swept Volume:
0.129 μL
Ask our AI about this Product
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
  • Absolute zero or low dead volume formats
  • Includes two stainless steel nuts and ferrules
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description0.020 in. through hole
IncludesUnion with 2 stainless steel nuts and ferrules
Swept Volume0.129 μL
For Use With (Equipment)Waters Fittings
MaterialStainless Steel
TypeUnion
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link