Search Thermo Fisher Scientific
Catalog Number | Description | Level |
---|---|---|
UR11001 | UrichemTRAK Controls, Assayed | 1 |
UR-MP | UrichemTRAK Controls, Assayed, Bi-level | Bi-level |
UR22002 | UrichemTRAK Controls, Assayed | 2 |
10014127 | UrichemTRAK Control, Vista Vials, Assayed, Bi-level | Bi-level |
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient Vista™ vial format. Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.