pcDNA™5/TO Mammalian Expression Vector
pcDNA™5/TO Mammalian Expression Vector
Gibco™
pcDNA™5/TO Mammalian Expression Vector
pcDNA5/TO is a 5.7 kb expression vector designed for use with the T-REx System (Cat. Nos. K1020-01 and K1020-02) andRead more
Have Questions?
Catalog number V103320
also known as V1033-20
Price (USD)/ 20 µg
882.65
Online exclusive
924.00 
Save 41.35 (4%)
-
Add to cart
Price (USD)/ 20 µg
882.65
Online exclusive
924.00 
Save 41.35 (4%)
Add to cart
pcDNA™5/TO Mammalian Expression Vector
Catalog numberV103320
Price (USD)/ 20 µg
882.65
Online exclusive
924.00 
Save 41.35 (4%)
-
Add to cart
pcDNA5/TO is a 5.7 kb expression vector designed for use with the T-REx System (Cat. Nos. K1020-01 and K1020-02) and Expi293 Inducible platform (Cat. Nos. A39251 and A39252). It utilizes the complete CMV promoter and adds control elements from the bacterial tetracycline resistance operon to effectively repress and derepress transcription from one of the strongest mammalian promoter sequences known.
The pcDNA5/TO vector allows tetracycline-regulated expression of the gene of interest in mammalian host cells expressing the Tet repressor (TetR) from the pcDNA6/TR vector (Catalog no. V1025-20).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemInducible
Delivery TypeTransfection
Inducing AgentTetracycline
PromoterCMV/TO
Product TypePichia Expression Vector
Selection Agent (Eukaryotic)Hygromycin
Protein TagUntagged
Cloning MethodRestriction Enzyme/MCS
Quantity20 μg
VectorpcDNA
Product LineExpi293™, T-REx™, pcDNA™
For Use With (Application)Regulated Expression
Unit Size20 µg
Contents & Storage
Store pcDNA5/TO at -20°C