Sensititre™ Vizion™ Digital MIC Viewing System
Sensititre™ Vizion™ Digital MIC Viewing System
Thermo Scientific™

Sensititre™ Vizion™ Digital MIC Viewing System

Consolidate all offline testing on a single instrument, for maximum efficiency, with the Thermo Scientific™ Sensititre™ Vizion™ Digital MIC Viewing System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
V2021Sensititre Vizion Instrument
V2020-SYESensititre Vizion System Plus SWIN Epidemiology Module
V2020-SYSSensititre Vizion System
Catalog number V2021
Price (USD)
-
Request A Quote
Description:
Sensititre Vizion Instrument
Ask our AI about this Product
Consolidate all offline testing on a single instrument, for maximum efficiency, with the Thermo Scientific™ Sensititre™ Vizion™ Digital MIC Viewing System, which combines automation with visual results. Just place your plate in the Vizion instrument, and customizable lighting options facilitate efficient, manual reading of all organisms. The easy-to-read digital plate image allows users to easily read and record MIC results, eliminating manual reading errors. Using the powerful Thermo Scientific™ Sensititre™ SWIN™ Software System, store images to expedite teaching, sharing, and decision-making, with LIS connectivity.
Using the powerful Thermo Scientific™ Sensititre™ SWIN™ Software System, store images with LIS connectivity to expedite teaching, sharing, and decision-making. Combining automation with visual results, the Sensititre Vizion Digital MIC Viewing System consolidates all offline testing on a single instrument for maximum efficiency.
  • Fully customizable Expert System and LIS connectivity save time, reduce errors and speed valuable information to clinicians
  • On-board quality control management modules facilitate recordkeeping
  • Epidemiology capabilities highlight emerging resistance patterns and provide easy annual report options

Sensititre Vizion System
  • V2020-SYS includes Vizion instrument and SWIN Computer
  • V2020-SYS-LIS includes Vizion instrument, SWIN Computer and LIS interface
  • V2020-SYE includes Vizion instrument, SWIN Computer and SWIN Epidemiology Module
  • V2020-SYE-LIS includes Vizion instrument, SWIN Computer, SWIN Epidemiology Module and LIS interface

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Antimicrobial AgentsVarious
Depth (English)13.9 in.
Depth (Metric)35.4 cm
DescriptionSensititre Vizion Instrument
Electrical Requirements220/240VAC; 60Hz; 8w
Height (English)12.4 in.
Height (Metric)31.5 cm
Regulatory StatusETL safety tested, CE marked, EMC tested, FDA-cleared
Weight (English)21 lb.
Weight (Metric)9.5 kg
Width (English)10.4 in.
Width (Metric)26.3 cm
Product TypeDigital MIC Viewing System
SpeciesMultiple
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach
Sensititre Pod Header

Explore our wide range of standard MIC plates and antimicrobials

Our latest Sensititre Plate Guides include everything you need to get to know the Sensititre System.

Download now ›

MBD-1103-PDP Pod-278x123

Thermo Scientific Sensititre System

Combine manual, semi-automated, and fully automated tests within a single integrated AST platform using broth microdilution (BMD), the gold standard for AST accuracy.¹

Explore the full workflow ›

¹Jayol A, Nordmann P, André C, Poirel L, Dubois V. Evaluation of three broth microdilution systems to determine colistin susceptibility of Gram-negative bacilli. J Antimicrob Chemother. 2018; 73: 1272-1278. PubMed
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Share catalog number, name or link

1x1 image pixel for data collection