Invitrogen™
pcDNA™6/myc-His A, B, & C Mammalian Expression Vectors
Catalog number: V22120
Have Questions?
Invitrogen™
pcDNA™6/myc-His A, B, & C Mammalian Expression Vectors
Catalog number: V22120
Have Questions?
Catalog Number
V22120
Unit Size
20 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
V2212020 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
All pcDNA™ vectors contain a strong promoter for high-level expression in mammalian cells, a choice of selection marker for generating stable cell lines, and an epitope tag for easy detection with a monoclonal antibody and rapid purification on nickel-chelating resin. Each vector is available in three reading frames to simplify cloning in-frame with the fusion tag.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible System
Constitutive
Delivery Type
Transfection
Promoter
CMV
Product Type
Mammalian Expression Vector
Selection Agent (Eukaryotic)
Blasticidin
Protein Tag
His Tag (6x), c-Myc Epitope Tag
Cloning Method
Restriction Enzyme/MCS
Vector
pcDNA
Product Line
pcDNA™
For Use With (Application)
Constitutive Expression
Contents & Storage
Vectors (and appropriate expression control) are supplied supercoiled and lyophilized. Stored at -20°C. All vectors are guaranteed stable for 6 months when properly stored.