Sensititre™ Complete Automated AST System
Sensititre™ Complete Automated AST System
Sensititre™ Complete Automated AST System
Sensititre™ Complete Automated AST System
Sensititre™ Complete Automated AST System
Sensititre™ Complete Automated AST System
Sensititre™ Complete Automated AST System
Thermo Scientific™
Sensititre™ Complete Automated AST System
Perform all susceptibility testing on a single platform utilizing the superior sensitivity of true MIC results with the Thermo Scientific™ Sensititre™ Complete Automated AST System.
Have Questions?
Catalog NumberDescription
V3000-VZSensititre Complete Automated AST System
Catalog number V3000-VZ
Price (USD)
-
Request A Quote
Description:
Sensititre Complete Automated AST System
Ask our AI about this Product
Perform all susceptibility testing on a single platform utilizing the superior sensitivity of true MIC results with the Thermo Scientific™ Sensititre™ Complete Automated AST System. The complete automated system includes the Sensititre™ ARIS™ HiQ™, OptiRead™, Sensititre AIM™, Sensititre™ Vizion™ System, Sensititre™ SWIN™ computer, Nephelometer, barcode printer and barcode labels.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionSensititre Complete Automated AST System
Product TypeSensititre Complete Automated System
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link