Vybrant™ Alexa Fluor™ 555 Lipid Raft Labeling Kit
Vybrant™ Alexa Fluor™ 555 Lipid Raft Labeling Kit
Invitrogen™
Vybrant™ Alexa Fluor™ 555 Lipid Raft Labeling Kit
Vybrant Lipid Raft Labeling Kits are designed to provide convenient, reliable and extremely bright fluorescent labeling of lipid rafts inRead more
Have Questions?
Catalog number V34404
Price (USD)/ 1 kit
711.65
Online exclusive
714.00 
Save 2.35
-
Add to cart
Price (USD)/ 1 kit
711.65
Online exclusive
714.00 
Save 2.35
Add to cart
Vybrant™ Alexa Fluor™ 555 Lipid Raft Labeling Kit
Catalog numberV34404
Price (USD)/ 1 kit
711.65
Online exclusive
714.00 
Save 2.35
-
Add to cart
Vybrant Lipid Raft Labeling Kits are designed to provide convenient, reliable and extremely bright fluorescent labeling of lipid rafts in live cells. Kits include cholera toxin subunit B (CT-B) labeled with either the green-fluorescent Alexa Fluor 488, the orange-fluorescent Alexa Fluor 555 or the red-fluorescent Alexa Fluor 594 dye, as well as an antibody that specifically recognizes CT-B. A 10X solution of phosphate-buffered saline (10X PBS), pH 7.2, and detailed instructions are also included in the kits.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Product LineAlexa Fluor™
Protein SubtypeCholera Toxin
Shipping ConditionWet Ice
Label TypeAlexa Fluor Dyes
Product TypeLipid Raft Labeling Kit
SubCellular LocalizationCell Membranes and Lipids
Unit Size1 kit
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.