Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Dead Cell Apoptosis Kit with Annexin V PE and SYTOX™ Green, for flow cytometry |
Kit Contents | Contains 1 vial of annexin V, R-PE conjugate (250 μL), 1 vial of SYTOX green stain (100 μL), and 1 bottle of annexin binding buffer (5X solution, 15 mL). |
Product Type | Apoptosis Detection Kit |
Flow Cytometer Laser Lines | 561, 488 |
Excitation/Emission | 503, 488/524, 575 |
No. of Reactions | 50 |
Shipping Condition | Wet Ice |
Conjugate | SYTOX Green, R-PE |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 1 kit |
Description | Dead Cell Apoptosis Kit with Annexin V Alexa Fluor 488 & Propidium Iodide (PI) |
Kit Contents | Contains 1 vial of annexin V, Alexa Fluor 488 conjugate (250 μL), 1 vial of propidium iodide (PI, 100 μL), and 1 bottle of annexin binding buffer (5X solution, 15 mL). |
Product Type | Apoptosis Detection Kit |
Flow Cytometer Laser Lines | 488 |
Excitation/Emission | 499, 535/521, 617 |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 488, Propidium Iodide |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 50 assays |
Description | Dead Cell Apoptosis Kit with Annexin V FITC and PI, for flow cytometry |
Kit Contents | Contains 1 vial of annexin V, FITC conjugate (250 μL ), 1 vial of propidium iodide (PI, 100 μL), and 1 bottle of annexin binding buffer (5X solution, 15 mL). |
Product Type | Apoptosis Detection Kit |
Flow Cytometer Laser Lines | 488 |
Excitation/Emission | 494, 535/518, 617 |
No. of Reactions | 50 |
Shipping Condition | Wet Ice |
Conjugate | FITC, Propidium Iodide |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 1 kit |
Description | Dead Cell Apoptosis Kit with Annexin V APC and SYTOX Green, for flow cytometry |
Kit Contents | Contains 1 vial of annexin V, APC conjugate (250 μL), 1 vial of SYTOX green stain (100 μL),and 1 bottle of annexin binding buffer (5X solution, 15 mL) . |
Product Type | Apoptosis Detection Kit |
Flow Cytometer Laser Lines | 633/635, 488 |
Excitation/Emission | 503, 650/524, 660 |
No. of Reactions | 50 |
Shipping Condition | Wet Ice |
Conjugate | SYTOX Green, APC |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 1 kit |
Description | Dead Cell Apoptosis Kit with Annexin V Alexa Fluor 488 & Propidium Iodide (PI) |
Kit Contents | Contains 5 vials of annexin V, Alexa Fluor 488 conjugate (250 μL per vial), 1 vial of propidium iodide (PI, 100 μL), and 1 bottle of annexin binding buffer (5X solution, 50 mL). |
Product Type | Apoptosis Detection Kit |
Flow Cytometer Laser Lines | 488 |
Excitation/Emission | 499, 535/521, 617 |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 488, Propidium Iodide |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 250 assays |
Catalog Number | Specifications | Unit Size | Conjugate | Excitation/Emission | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
V35112 | 1 kit | SYTOX Green, R-PE | 503, 488/524, 575 | 638.65 Online price 694.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
V13241 | 50 assays | Alexa Fluor 488, Propidium Iodide | 499, 535/521, 617 | 205.65 Online price 219.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
V13242 | 1 kit | FITC, Propidium Iodide | 494, 535/518, 617 | 308.65 Online price 332.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
V35113 | 1 kit | SYTOX Green, APC | 503, 650/524, 660 | 634.65 Online price 662.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
V13245 | 250 assays | Alexa Fluor 488, Propidium Iodide | 499, 535/521, 617 | 691.65 Online price 752.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|