Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Vybrant FAM Caspase-8 Assay Kit |
Format | Tube |
Kit Contents | 1 vial of FAM-LETD-FMK caspase-8 reagent (FLICA reagent), 1 vial Hoechst 33342, 1 vial of propidium iodide, 1 vial of DMSO, 1 bottle of apoptosis fixative solution, and 1 bottle of apoptosis wash buffer (10X solution). |
Product Type | Caspase Assay Kit |
Flow Cytometer Laser Lines | UV, 488 |
Excitation/Emission | FAM-LETD-FMDK: 488/530 PI: 535/617 Hoechst 33342: 350/461 |
Shipping Condition | Room Temperature |
Product Line | Vybrant |
Detection Method | Fluorescence |
For Use With (Equipment) | Flow Cytometer |
Label or Dye | FAM, propidium iodide, Hoechst 33342 |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in refrigerator (2–8°C) and protect from light. |
Unit Size | 25 assays |
Description | Vybrant FAM Poly Caspases Assay Kit |
Format | Tube |
Kit Contents | 1 vial of FAM-VAD-FMK poly caspases reagent (FLICA reagent), 1 vial Hoechst 33342, 1 vial of propidium iodide, 1 vial of DMSO, 1 bottle of apoptosis fixative solution, and 1 bottle of apoptosis wash buffer (10X solution) |
Product Type | Caspase Assay Kit |
Flow Cytometer Laser Lines | UV, 488 |
Excitation/Emission | FAM-VAD-FMDK: 488/530 PI: 535/617 Hoechst 33342: 350/461 |
Shipping Condition | Room Temperature |
Product Line | Vybrant |
Detection Method | Fluorescence |
For Use With (Equipment) | Flow Cytometer |
Label or Dye | FMK, Hoechst 33342, propidium iodide |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in refrigerator (2–8°C) and protect from light. |
Unit Size | 25 assays |
Description | Vybrant FAM Caspase-3 and -7 Assay Kit |
Format | Tube |
Kit Contents | 1 vial FAM-DEVD-FMK caspase-3 and -7 reagent (FLICA reagent, lyophilized solid), 1 vial of Hoechst 33342 (400 μL, 1 mM in water), 1 vial of PI (1 mL, 250 μg/mL in water), 1 vial of DMSO (0.5 mL), 1 bottle of apoptosis fixative solution (10% formaldehyde with methanol in PBS, 6 mL), and 1 bottle of 10x apoptosis wash buffer. |
Product Type | Caspase Assay Kit |
Flow Cytometer Laser Lines | UV, 488 |
Excitation/Emission | FAM-DEVD-FMD: 388/530 PI: 535/617 Hoechst 33342: 350/461 |
Shipping Condition | Room Temperature |
Product Line | Vybrant |
Detection Method | Fluorescence |
For Use With (Equipment) | Flow Cytometer |
Label or Dye | FAM, Hoechst 33342, propidium iodide |
Label Type | Other Label(s) or Dye(s) |
Storage Requirements | Store in refrigerator (2–8°C) and protect from light. |
Unit Size | 25 assays |
Catalog Number | Specifications | Unit Size | Label or Dye | Description | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
V35119 | 25 assays | FAM, propidium iodide, Hoechst 33342 | Vybrant FAM Caspase-8 Assay Kit | 302.65 Online price 308.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
V35117 | 25 assays | FMK, Hoechst 33342, propidium iodide | Vybrant FAM Poly Caspases Assay Kit | 306.65 Online price 313.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
V35118 | 25 assays | FAM, Hoechst 33342, propidium iodide | Vybrant FAM Caspase-3 and -7 Assay Kit | 304.65 Online price 312.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|