pLenti6/UbC/V5-DEST™ Gateway™ Vector
Product Image
Invitrogen™

pLenti6/UbC/V5-DEST™ Gateway™ Vector

Invitrogen™'s pLenti6⁄UbC⁄V5-DEST™ Gateway™ Vector Kit is part of our ViraPower™ UbC Lentiviral Gateway™ Expression Kit (catalog K4990-00). The pLenti6⁄UbC⁄V5-DEST™ Gateway™Read more
Have Questions?
Catalog number V49910
also known as V499-10
Price (USD)/ 6 µg
2,380.00
-
Add to cart
Price (USD)/ 6 µg
2,380.00
Add to cart
pLenti6/UbC/V5-DEST™ Gateway™ Vector
Catalog numberV49910
Price (USD)/ 6 µg
2,380.00
-
Add to cart
Invitrogen™'s pLenti6⁄UbC⁄V5-DEST™ Gateway™ Vector Kit is part of our ViraPower™ UbC Lentiviral Gateway™ Expression Kit (catalog K4990-00).

The pLenti6⁄UbC⁄V5-DEST™ Gateway™ Vector is a Gateway™-adapted ViraPower™ lentiviral expression vector for lentiviral-based expression of a target gene in dividing and non-dividing mammalian cells. The vector has the UbC promoter for driving constitutive but physiological levels of expression of the target gene and the blasticidin selection marker for stable selection in mammalian cells.

Advantages
• Stable expression
• Long-term experiments
• Accurate titer of functional virus
• Flexible and versatile Gateway™ recombination cloning technology

Key Features
• UbC promoter
• V5 epitope tag at C terminus
• Blasticidin selection

Kit includes
• A lacZ vector as a positive control, pLenti6⁄UbC⁄V5-GW⁄lacZ
• Stbl3™ competent cells

For research use only. Not intended for any therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeLentiviral
PromoterUbC
Product TypeLentiviral Expression Vector
Selection Agent (Eukaryotic)Blasticidin
Protein TagV5 Epitope Tag
Cloning MethodGateway
Quantity6 μg
VectorpDEST, pLenti
Product LineGateway™, ViraPower™
For Use With (Application)Viral Expression
Unit Size6 µg
Contents & Storage
Vector Kit (Box 1)
pLenti6⁄UbC⁄V5-DEST™:6 μg, store at -20°C
pLenti6⁄UbC⁄V5-GW⁄lacZ Control vector: 10 ug, store at -20°C

One Shot™ Stbl3™ Chemically Competent E. coli(Box 2)
Stbl3™ Cells: 21 x 50 μL, store at -80°C
pUC19 control DNA (10 pg⁄μL): 50 μL, store at -80°C
S.O.C. Medium: 6 mL, store at -80°C