Invitrogen™
pEXP2-DEST Vector Kit
Catalog number: V96002
Have Questions?
Invitrogen™
pEXP2-DEST Vector Kit
Catalog number: V96002
Have Questions?
Catalog Number
V96002
also known as V960-02
Unit Size
6 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
V96002
also known as V960-02
6 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
To fit all of your expression needs, Invitrogen offers state-of-the-art Gateway™ destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins. All Gateway™ destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is an entry clone or an Ultimate™ RF Clone. The following table lists the wide range of destination vectors available.
Additional materials required, available separately: Gateway™ entry clone, Gateway™ LR Clonase™ enzyme mix, and reaction buffer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Promoter
T7
Product Type
Gateway System Destination Expression Vector
Selection Agent (Eukaryotic)
None
Antibiotic Resistance Bacterial
Ampicillin (AmpR), Zeocin™ (ZeoR)
Protein Tag
His Tag (6x), V5 Epitope Tag
Cloning Method
Gateway
Vector
pEXP, pDEST
Product Line
Expressway™, Gateway™
Contents & Storage
All destination vectors are provided lyophilized and supercoiled.