Search Thermo Fisher Scientific
Catalog Number | For Use With (Equipment) |
---|---|
VALPRONIR | RESULT and Antaris NIR spectrometers |
Achieve cGMP and FDA regulatory compliance with the Thermo Scientific™ ValPro™ System Qualification Software for NIR Analyzers. Providing a full set of tools for DQ, IQ, OQ and PQ testing of the Thermo Scientific™ Antaris™ FT-NIR Analyzers, this package includes software, extensive documentation and a validation wheel with NIST™ traceable and USP-recommended standards.
The ValPro System Qualification Software for NIR Analyzers Features:
Share catalog number, name or link