Thermo Scientific™

Vanquish™ Fraction Collector

Catalog number: VF-F20-A-01
Have Questions?
Thermo Scientific™

Vanquish™ Fraction Collector

Catalog number: VF-F20-A-01
Have Questions?
Experience compelling fraction purity with the Thermo Scientific™ Vanquish™ Fraction Collector (VFC) using innovative valve technology for enhanced resolution, product recovery, and minimized carry-over. The FC can be used with any analytical flow Thermo Scientific™ Vanquish™ HPLC and UHPLC system, any detector including MS, all under Thermo Scientific™ Chromeleon™ Chromatography Data System control. The VFC is the ideal addition to your Vanquish LC system, combining the utmost application flexibility with high-performance fractionation options to preserve superior separation within your fractionation process and protecting the sample integrity.
 
Catalog Number
VF-F20-A-01
Unit Size
Each
Description
Integral Fraction Collector FT: Thermostatted fraction collector for integration into Vanquish LC stacks
For Use With (Equipment)
Vanquish HPLC and UHPLC Systems
Type
Integrated Fraction Collector
Price (USD)
Full specifications
TypeIntegrated Fraction Collector
Biocompatible VersionYes, pH range 2-12, chloride concentration up to 1 mol/L
Fraction Capacity4 segments useable for sample racks or well plates with SBS footprint format
CarryoverCarry over < 0.15% with flush enabled
Collection ModesPeak-based, time-based and manual fractionation; pooling, wrapping, concatenating
DescriptionIntegral Fraction Collector FT: Thermostatted fraction collector for integration into Vanquish LC stacks
Flow Rate Range0.05 mL/min - 10 mL/min
For Use With (Application)Fractionation and purification
For Use With (Equipment)Vanquish HPLC and UHPLC Systems
Fraction Compartment Temperature Range4-40°C, cooling ≥ 23 K below ambient at < 80% relative humidity
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionFor Use With (Equipment)TypePrice (USD)
VF-F20-A-01Full specifications
EachIntegral Fraction Collector FT: Thermostatted fraction collector for integration into Vanquish LC stacksVanquish HPLC and UHPLC SystemsIntegrated Fraction CollectorRequest A Quote
TypeIntegrated Fraction Collector
Biocompatible VersionYes, pH range 2-12, chloride concentration up to 1 mol/L
Fraction Capacity4 segments useable for sample racks or well plates with SBS footprint format
CarryoverCarry over < 0.15% with flush enabled
Collection ModesPeak-based, time-based and manual fractionation; pooling, wrapping, concatenating
DescriptionIntegral Fraction Collector FT: Thermostatted fraction collector for integration into Vanquish LC stacks
Flow Rate Range0.05 mL/min - 10 mL/min
For Use With (Application)Fractionation and purification
For Use With (Equipment)Vanquish HPLC and UHPLC Systems
Fraction Compartment Temperature Range4-40°C, cooling ≥ 23 K below ambient at < 80% relative humidity
Unit SizeEach
Showing 1 of 1

Vanquish Fraction Collector features:

  • Fully integrated fraction collector in Vanquish Core HPLC, Vanquish Flex UHPLC, and Vanquish Horizon UHPLC systems for superior user experience
  • Broad flow range from 0.05 – 10 mL/min for wide application flexibility
  • Maximized recovery of the purest fractions through wash and flush procedures for high precision and accuracy
  • Preserved compound resolution through minimized dispersion, low carry-over, fast fractionation times, and adaptable fraction collection modes
  • Optimal system setup with Thermo Scientific™ Viper™ Fingertight Fitting Systems and automated delay volume determination
  • Controlled sample storage through stable thermostatting and protection against dust and light to preserve sample integrity
  • Advanced sample security through automated vial rack and well plate recognition via barcode reading
  • Precise needle positioning supporting a wide variety of sample containers and closures, including septum piercing
  • Confident biological sample processing through biocompatibe flow path
  • Advanced software features enable smooth integration
  • Commonly used for isolation of impurities or purification of active substances, like antibody, proteins, protein fragments, oligonucleotides, mRNA reagents, traditional medicines and natural products.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text