Search Thermo Fisher Scientific
Catalog Number | Description | Weight (Metric) | PC Connection |
---|---|---|---|
VH-A90-A | Barcode reading: <ul> <li>Empty shelf level detection</li> <li>Rack/well plate verification</li> <li>Inventory management</li> </ul> | <35 kg | USB 2.0 |
Employ an intelligent robotic that is fully integrated into the Thermo Scientific™ Vanquish™ UHPLC system with the Vanquish Charger module. With a capacity of 8832 samples (23 well plates), the Charger module automates sample loading into the Vanquish system for extra-long unattended operation. Environmental controls ensure reliable sample protection. A barcode reader automatically inventories each plate or rack within the device every time the door is closed, allowing users to add samples at anytime even while the Vanquish UHPLC system is in operation.
Find more HPLC and UHPLC instruments, components, related products, services and resources.
Share catalog number, name or link