Thermo Scientific™

Vanquish™ Neo UHPLC System

Catalog number: VN-S10-A-01
Have Questions?
Thermo Scientific™

Vanquish™ Neo UHPLC System

Catalog number: VN-S10-A-01
Have Questions?

For researchers pursuing the next scientific breakthrough, the Thermo Scientific™ Vanquish™ Neo UHPLC system delivers maximum performance 24/7 for nano-, capillary-, and micro-flow LC-MS. Thermo Scientific™ ProFlow™ XR 1500 bar pump technology ensures excellent retention time precision from nano-flow up to 100 μL/min, the unique low-flow split-loop autosampler enhances sample throughput and injection performance, and integrated system intelligence streamlines method editing and simplifies operation. From -omics and biopharma to high-throughput translational research, the Vanquish Neo UHPLC system is the new standard in low-flow LC.


View Vanquish Horizon UHPLC resources | Product tour | Build your HPLC | HPLC Support Center

 
Catalog Number
VN-S10-A-01
Unit Size
Each
Description
Vanquish Neo UHPLC System
Type
Low-Flow Liquid Chromatography System
Price (USD)
Full specifications
TypeLow-Flow Liquid Chromatography System
Column Temperature Range5°C above ambient to 80°C (depending on configuration)
Max. Pressure1500 bar (21,750 psi)
DescriptionVanquish Neo UHPLC System
Flow Rate Range1 nL/min - 100 μL/min, 1 nL increments (100 nL/min - 100 μL/min, recommended)
Dimensions (L x W x H)740 x 450 x 650 mm
Green FeaturesLess waste, more energy efficient and responsibly packaged
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionTypePrice (USD)
VN-S10-A-01Full specifications
EachVanquish Neo UHPLC SystemLow-Flow Liquid Chromatography SystemRequest A Quote
TypeLow-Flow Liquid Chromatography System
Column Temperature Range5°C above ambient to 80°C (depending on configuration)
Max. Pressure1500 bar (21,750 psi)
DescriptionVanquish Neo UHPLC System
Flow Rate Range1 nL/min - 100 μL/min, 1 nL increments (100 nL/min - 100 μL/min, recommended)
Dimensions (L x W x H)740 x 450 x 650 mm
Green FeaturesLess waste, more energy efficient and responsibly packaged
Unit SizeEach
Showing 1 of 1
All-in-one nano-, capillary-, and micro-flow LC for high-sensitivity LC-MS workflows
  • ProFlow technology enables active flow control for excellent retention time precision
  • Vial bottom detection technology delivers precise and accurate injections from limited sample volumes, decreasing sample waste
  • Automated multi-wash procedures minimize system carryover for improved low-level analyte sensitivity
Accelerating productivity with long-term, trouble-free analysis
  • Unique, 1500 bar split-loop autosampler design minimizes sample dispersion and system maintenance while enhancing workflow versatility
  • Fast sample loading and equilibration at maximum system pressure improves sample throughput
  • Next-generation Thermo Scientific™ SmartInject technology improves retention time precision, column lifetime, and electrospray ionization stability
Enabling non-chromatographers and LC-MS experts to get high-quality results
  • The Thermo Scientific Vanquish User Interface provides intuitive at-system and remote control, system status monitoring, standalone diagnostics, and guided maintenance routines
  • Integrated system intelligence simplifies daily operation and guides the creation of optimal LC methods
Seamless LC-MS integration

The Vanquish Neo UHPLC System seamlessly integrates with Thermo Scientific mass spectrometers through a comprehensive suite of ESI sources. Harness speed, robustness and sensitivity with triple quadrupole MS systems or resolution, mass accuracy, and versatility with Thermo Scientific™ Orbitrap™ MS systems.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text