Vanquish™ High-throughput LC systems
Vanquish™ High-throughput LC systems
Vanquish™ High-throughput LC systems
Vanquish™ High-throughput LC systems
Thermo Scientific™

Vanquish™ High-throughput LC systems

Green features

Vanquish High-throughput LC systems can boost your lab's productivity by increasing throughput and reducing cost per sample without added bench space. Vanquish Duo UHPLC Systems are an ACT labeled product with a My Green Lab ACT Ecolabel.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
VQDUO-DUALLCVanquish Duo UHPLC Systems
VQDUO-TANDEMVanquish Tandem LC Systems
Catalog number VQDUO-DUALLC
Price (USD)
-
Description:
Vanquish Duo UHPLC Systems

Use the Thermo Scientific™ Vanquish™ High-throughput LC Systems to improve productivity and save bench space. Vanquish Tandem LC systems increase sample throughput and MS utilization for LC-MS applications by eliminating column washing and equilibration steps from method cycle time. Vanquish Duo UHPLC systems combine two completely independent LC flow paths to double sample throughput or run independent analyses in parallel. In addition, Vanquish High-throughput LC Systems expand the benefits of the Vanquish platform without compromising performance, robustness, or ease-of-use.

View Vanquish HPLC resourcesHPLC Support Center

Two flow paths. One integrated system.

  • Increase your sample throughput
  • Free up valuable bench space
  • Enhance qualitative and quantitative sample knowledge
  • Reduce your cost per sample
  • Streamline your workflow using intelligent software tools

Vanquish Duo UHPLC systems
Double your throughput, deepen your sample knowledge, and better utilize your bench space with the Vanquish Duo system, a two channel LC for improved return on investment.

Vanquish Tandem LC systems
Increase your lab's throughput and return on investment for LC-MS assays with the Vanquish Tandem LC system for offline column regeneration.

More productivity with ease

  • Save time spent on user training with Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) wizards for creating Vanquish High-throughput system methods.
  • Control two independent flow paths from a single user interface with fully compliant Chromeleon CDS.
  • Save time using the Chromeleon CDS software for data analysis.
  • Seamlessly integrate with a mass spectrometry or other detector.

Sustainability

The Vanquish Duo UHPLC Systems are an ACT labeled product with a My Green Lab ACT Ecolabel.

The My Green Lab ACT Ecolabel serves as the premier ecolabel for lab products, encompassing consumables, chemicals, and equipment. Standing for Accountability, Consistency, and Transparency (ACT), this certification offers clear, standardized information about a product's environmental footprint across its life cycle covering manufacturing, energy and water use, packaging, and end-of-life disposal. Leverage the My Green Lab ACT Ecolabel to make informed purchasing choices based on transparent environmental data so you can advance sustainability without sacrificing performance. Learn more about the ACT Ecolabel and participating Thermo Scientific products here.

Specifications
DescriptionVanquish Duo UHPLC Systems
For Use With (Application)Two channel LC doubles throughput, deepens sample knowledge, saves on bench space, and improves return on investment.
Control the system with easy to use Chromeleon CDS.
Green FeaturesACT Ecolabel (Platinum)
For Use With (Equipment)Vanquish Horizon or Vanquish Flex Modules
Max. Pressure150 MPa, depending on configuration
Solvent Channels2
TypeHPLC or Biocompatible UHPLC
Unit SizeEach