Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Get high accuracy, premium performance and ultimate flexibility with the Thermo Scientific™ Orion™ Versa Star Pro™ Benchtop pH Meter. Meet your most challenging applications for pH, mV, ORP and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. Perform up to a 6 point pH calibration, log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer. Use the included electrode stand to easily place sensors in samples.
Accuracy (Temperature) | ±0.1°C |
Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
Accuracy (pH) | ±0.002 |
Data Management Details | Transfer single, range or all data points to printer or computer |
Description | Orion Versa Star Pro pH benchtop meter TRIS kit with TRIS-compatible ROSS Ultra pH/ATC electrode |
Outputs | USB, RS-232 |
Includes | Versa Star Pro meter with pH module, 8302BNUMD ROSS Ultra glass-body pH/ATC electrode, 096019 stirrer probe, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
Measurement Modes | pH, mV, Relative mV, ORP with Temperature |
Calibration | pH with calibration editing option, relative mV, ORP and temperature |
Range (pH) | -2 to 20 |
Resolution (pH) | 0.001, 0.01, 0.1 |
Range (mV) | ±2000.0 |
Range (ORP) | ±2000.0mV |
Resolution (mV) | 0.1mV |
Calibration Points | 1 to 6 |
Temperature Range (Metric) | -5°C to +105°C |
Temperature Range (English) | 22.0°F to 221.0°F |
Resolution (Temperature) | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Temperature Probe Calibration | 1 point temperature offset calibration |
Display Type | LCD, Color Graphic with User-Customizable Options |
Backlight Option | Yes |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Data Points | 2000 with Date and Time Stamp |
Log Function Type | Automatic with Auto-Read, Timed and Single Shot Measure Modes; Manual |
Alarm Output | Set point alarm, high/low limit alarm, calibration due alarm |
Inputs | BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe) |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
IP Rating | IP-54 |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) |
Dimensions (L x W x H) | 28 cm x 24 cm x 10 cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2 cm (0.8 in.) L & H |
Weight (Metric) | 1.2 kg, modules add 0.2 kg each |
Weight (English) | 2.5 lb., modules add 0.5 lb. each |
Warranty | 3 Year Meter and Module Warranty |
Keypad | Numeric with menu-specific function keys and dual purpose scroll/shortcut keys |
Methods | 10 per channel |
Power Rating | DC input: 9VDC, 1A |
Power Supply | Universal AC adapter (included) |
Probe Type | pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8302BNUMD ROSS Ultra Triode pH/ATC electrode, 096019 Orion Star stirrer probe) |
Languages | English, Spanish, French, Italian, German, Chinese, Portuguese and Korean |
Range (Relative mV) | ±2000.0mV |
Relative Humidity Range | 5 to 85%, non-condensing |
Isopotential Point | 7.000pH (adjustable) |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Channels | 4 - customizable with modules |
Unit Size | Each |
Accuracy (Temperature) | ±0.1°C |
Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
Accuracy (pH) | ±0.002 |
Data Management Details | Transfer single, range or all data points to printer or computer |
Description | Orion Versa Star Pro pH benchtop meter with stand |
Outputs | USB, RS-232 |
Includes | Versa Star Pro meter with pH module, electrode stand, 100-240V universal power adapter, computer cable |
Measurement Modes | pH, mV, Relative mV, ORP with Temperature |
Calibration | pH with calibration editing option, relative mV, ORP and temperature |
Range (pH) | -2 to 20 |
Resolution (pH) | 0.001, 0.01, 0.1 |
Range (mV) | ±2000.0 |
Range (ORP) | ±2000.0mV |
Resolution (mV) | 0.1mV |
Calibration Points | 1 to 6 |
Temperature Range (Metric) | -5°C to +105°C |
Temperature Range (English) | 22.0°F to 221.0°F |
Resolution (Temperature) | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Temperature Probe Calibration | 1 point temperature offset calibration |
Display Type | LCD, Color Graphic with User-Customizable Options |
Backlight Option | Yes |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Data Points | 2000 with Date and Time Stamp |
Log Function Type | Automatic with Auto-Read, Timed and Single Shot Measure Modes; Manual |
Alarm Output | Set point alarm, high/low limit alarm, calibration due alarm |
Inputs | BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe) |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
IP Rating | IP-54 |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) |
Dimensions (L x W x H) | 28 cm x 24 cm x 10 cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2 cm (0.8 in.) L & H |
Weight (Metric) | 1.2 kg, modules add 0.2 kg each |
Weight (English) | 2.5 lb., modules add 0.5 lb. each |
Warranty | 3 Year Meter and Module Warranty |
Keypad | Numeric with menu-specific function keys and dual purpose scroll/shortcut keys |
Methods | 10 per channel |
Power Rating | DC input: 9VDC, 1A |
Power Supply | Universal AC adapter (included) |
Probe Type | pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe, stirrer probe (purchase separately) |
Languages | English, Spanish, French, Italian, German, Chinese, Portuguese and Korean |
Range (Relative mV) | ±2000.0mV |
Relative Humidity Range | 5 to 85%, non-condensing |
Isopotential Point | 7.000pH (adjustable) |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Channels | 4 - customizable with modules |
Unit Size | Each |
Accuracy (Temperature) | ±0.1°C |
Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
Accuracy (pH) | ±0.002 |
Data Management Details | Transfer single, range or all data points to printer or computer |
Description | Orion Versa Star Pro pH benchtop meter difficult sample kit with ROSS Sure-Flow pH electrode and ATC probe |
Outputs | USB, RS-232 |
Includes | Versa Star Pro meter with pH module, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD stainless steel ATC probe, 096019 stirrer probe, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable |
Measurement Modes | pH, mV, Relative mV, ORP with Temperature |
Calibration | pH with calibration editing option, relative mV, ORP and temperature |
Range (pH) | -2 to 20 |
Resolution (pH) | 0.001, 0.01, 0.1 |
Range (mV) | ±2000 |
Range (ORP) | ±2000.0mV |
Resolution (mV) | 0.1mV |
Calibration Points | 1 to 6 |
Temperature Range (Metric) | -5°C to +105°C |
Temperature Range (English) | 22.0°F to 221.0°F |
Resolution (Temperature) | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Temperature Probe Calibration | 1 point temperature offset calibration |
Display Type | LCD, Color Graphic with User-Customizable Options |
Backlight Option | Yes |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Data Points | 2000 with Date and Time Stamp |
Log Function Type | Automatic with Auto-Read, Timed and Single Shot Measure Modes; Manual |
Alarm Output | Set point alarm, high/low limit alarm, calibration due alarm |
Inputs | BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe) |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
IP Rating | IP-54 |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) |
Dimensions (L x W x H) | 28 x 24 x 10 cm (11 x 9.5 x 3.9 in.), modules add ≤ 2 cm (0.8 in.) L & H |
Weight (Metric) | 1.2 kg, modules add 0.2 kg each |
Weight (English) | 2.5 lb., modules add 0.5 lb. each |
Keypad | Numeric with menu-specific function keys and dual purpose scroll/shortcut keys |
Methods | 10 per channel |
Power Rating | DC input: 9VDC, 1A |
Power Supply | Universal AC adapter (included) |
Probe Type | pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe, stirrer probe |
Languages | English, Spanish, French, Italian, German, Chinese, Portuguese and Korean |
Range (Relative mV) | ±2000.0mV |
Relative Humidity Range | 5 to 85%, non-condensing |
Isopotential Point | 7.000pH (adjustable) |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Channels | 4 - customizable with modules |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Includes | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
VSTAR12 | Each | Orion Versa Star Pro pH benchtop meter TRIS kit with TRIS-compatible ROSS Ultra pH/ATC electrode | Versa Star Pro meter with pH module, 8302BNUMD ROSS Ultra glass-body pH/ATC electrode, 096019 stirrer probe, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable | 1,926.00 Online price 2,140.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
VSTAR10 | Each | Orion Versa Star Pro pH benchtop meter with stand | Versa Star Pro meter with pH module, electrode stand, 100-240V universal power adapter, computer cable | 1,710.00 Online price 1,900.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
VSTAR13 | Each | Orion Versa Star Pro pH benchtop meter difficult sample kit with ROSS Sure-Flow pH electrode and ATC probe | Versa Star Pro meter with pH module, 8172BNWP ROSS Sure-Flow glass-body pH electrode, 927007MD stainless steel ATC probe, 096019 stirrer probe, 810199 pH buffer kit, electrode stand, 100-240V universal power adapter, computer cable | 2,331.00 Online price 2,590.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
System Access Feature