Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Get high accuracy, premium performance and ultimate flexibility with the Thermo Scientific™ Orion™ Versa Star Pro™ pH/ISE Benchtop Multiparameter Meter. Meet your most challenging applications for pH, ion concentration, mV, ORP and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. Perform up to 6 point pH and ISE calibrations, log up to 2000 data point sets with time/date stamp and transfer calibration and data logs via USB or RS232 to a printer or computer. Select systems with one or two pH/ISE modules.
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) |
Outputs | USB (2), RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | Set point alarm, high/low limit alarm, calibration due alarm |
Backlight Option | Yes |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 6 |
Calibration Points (pH) | 1 to 6 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Versa Star Pro benchtop meter kit with two pH/ISE/mV modules, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and two stands |
Channels | 4 - customizable with modules |
Display Type | Color graphic LCD with user-customizable options |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 4 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Versa Star Pro meter, two VSTAR-ISE pH/ISE modules, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
Inputs | Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe) |
Isopotential Point | 7.000pH (adjustable), 1.000 ISE (adjustable) |
Keypad | Numeric with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German, Chinese, Portuguese and Korean |
Log Function Type | Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel |
Power Rating | DC input: 9VDC, 1A |
Power Supply | Universal AC adapter (included) |
Probe Type | pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe |
Range (ISE) | 0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 year meter and module warranty |
Dimensions (L x W x H) | 28cm x 24cm x 10cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H |
Weight (English) | 2.5 lb., modules add 0.5 lb. each |
Weight (Metric) | 1.2 kg, modules add 0.2 kg each |
Unit Size | Each |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) |
Outputs | USB (2), RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | Set point alarm, high/low limit alarm, calibration due alarm |
Backlight Option | Yes |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 6 |
Calibration Points (pH) | 1 to 6 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Versa Star Pro benchtop meter with pH/ISE/mV module and stand |
Channels | 4 - customizable with modules |
Display Type | Color graphic LCD with user-customizable options |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 4 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Versa Star Pro meter, VSTAR-ISE pH/ISE module, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
Inputs | BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe) |
Isopotential Point | 7.000pH (adjustable), 1.000 ISE (adjustable) |
Keypad | Numeric with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German, Chinese, Portuguese and Korean |
Log Function Type | Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel |
Power Rating | DC input: 9VDC, 1A |
Power Supply | Universal AC adapter (included) |
Probe Type | pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (purchase separately) |
Range (ISE) | 0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 year meter and module warranty |
Dimensions (L x W x H) | 28cm x 24cm x 10cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H |
Weight (English) | 2.5 lb., modules add 0.5 lb. each |
Weight (Metric) | 1.2 kg, modules add 0.2 kg each |
Unit Size | Each |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) |
Outputs | USB (2), RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | Set point alarm, high/low limit alarm, calibration due alarm |
Backlight Option | Yes |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 6 |
Calibration Points (pH) | 1 to 6 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Versa Star Pro benchtop meter kit with pH/ISE/mV module, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and stand |
Channels | 4 - customizable with modules |
Display Type | Color graphic LCD with user-customizable options |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 4 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Versa Star Pro meter, VSTAR-ISE pH/ISE module, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
Inputs | BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe) |
Isopotential Point | 7.000pH (adjustable), 1.000 ISE (adjustable) |
Keypad | Numeric with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German, Chinese, Portuguese and Korean |
Log Function Type | Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel |
Power Rating | DC input: 9VDC, 1A |
Power Supply | Universal AC adapter (included) |
Probe Type | pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe |
Range (ISE) | 0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 year meter and module warranty |
Dimensions (L x W x H) | 28cm x 24cm x 10cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H |
Weight (English) | 2.5 lb., modules add 0.5 lb. each |
Weight (Metric) | 1.2 kg, modules add 0.2 kg each |
Unit Size | Each |
AC Adapter | Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR) |
Outputs | USB (2), RS232 |
Accuracy (ISE) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (mV) | ±0.2 mV or ±0.05% of reading whichever is greater |
Accuracy (pH) | ±0.002 |
Advanced ISE Functions | Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stabilit |
Alarm Output | Set point alarm, high/low limit alarm, calibration due alarm |
Backlight Option | Yes |
Calibration | pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ |
Calibration Points | 1 point relative mV (RmV) mode, 1 point ORP mode |
Calibration Points (ISE) | 1 to 6 |
Calibration Points (pH) | 1 to 6 |
Certifications/Compliance | CE, TUV 3-1, FCC Class A |
Data Management Details | Transfer single, range or all data points to printer or computer |
Data Points | 2000 with date and time stamp |
Description | Orion Versa Star Pro benchtop meter with two pH/ISE/mV modules and stand |
Channels | 4 - customizable with modules |
Display Type | Color graphic LCD with user-customizable options |
IP Rating | IP 54 |
Measurement Modes | pH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature |
Range (mV) | ±2000.0 mV |
Range (pH) | -2.000 to 20.000 |
Resolution (ISE) | 0.0001 minimum, 1 to 4 significant digits (user selectable) |
Resolution (pH) | 0.1/0.01/0.001 |
Temperature Range (Metric) | -5.0°C to +105.0°C |
Includes | Versa Star Pro meter, two VSTAR-ISE pH/ISE modules, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate |
Inputs | Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe) |
Isopotential Point | 7.000pH (adjustable), 1.000 ISE (adjustable) |
Keypad | Numeric with menu-specific function keys and dual purpose scroll/shortcut keys |
Languages | English, Spanish, French, Italian, German, Chinese, Portuguese and Korean |
Log Function Type | Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with |
Memory | Non-volatile memory preserves data log, calibration log and meter settings |
Methods | 10 per channel |
Power Rating | DC input: 9VDC, 1A |
Power Supply | Universal AC adapter (included) |
Probe Type | pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (purchase separately) |
Range (ISE) | 0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none |
Range (ORP) | ±2000.0 mV |
Range (Relative mV) | ±2000.0 mV |
Relative Humidity Range | 5 to 85 %, non-condensing |
Resolution (mV) | 0.1mV |
Stability | Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting |
Temperature Accuracy | ±0.1°C |
Temperature (English) Operating | 41°F to 113°F (ambient) |
Temperature (Metric) Operating | 5°C to 45°C (ambient) |
Temperature Probe Calibration | 1 point temperature offset calibration |
Temperature Range (English) | 22.0°F to 221.0° F |
Temperature Resolution | 0.1°C, 0.1°F |
Temperature Selection | Manual or automatic with ATC temperature probe |
Warranty | 3 year meter and module warranty |
Dimensions (L x W x H) | 28cm x 24cm x 10cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H |
Weight (English) | 2.5 lb., modules add 0.5 lb. each |
Weight (Metric) | 1.2 kg, modules add 0.2 kg each |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Includes | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
VSTAR40B2 | Each | Orion Versa Star Pro benchtop meter kit with two pH/ISE/mV modules, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and two stands | Versa Star Pro meter, two VSTAR-ISE pH/ISE modules, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate | 3,384.00 Online price 3,760.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
VSTAR40A | Each | Orion Versa Star Pro benchtop meter with pH/ISE/mV module and stand | Versa Star Pro meter, VSTAR-ISE pH/ISE module, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate | 2,376.00 Online price 2,640.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
VSTAR40A2 | Each | Orion Versa Star Pro benchtop meter kit with pH/ISE/mV module, ROSS Ultra glass-body pH electrode, ATC probe, stirrer probe, solutions and stand | Versa Star Pro meter, VSTAR-ISE pH/ISE module, 8102BNUWP ROSS Ultra glass-body pH electrode, 927007MD Orion stainless steel ATC probe, 096019 Orion Star stirrer probe, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate | 2,916.00 Online price 3,240.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
VSTAR40B | Each | Orion Versa Star Pro benchtop meter with two pH/ISE/mV modules and stand | Versa Star Pro meter, two VSTAR-ISE pH/ISE modules, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate | 2,745.00 Online price 3,050.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
System Access Feature
Orion Versa Star Pro meters offer a system access feature that is designed to help ensure data integrity by preventing undesired access to various meter functions including date/time setting, data log and calibration log. The System Access feature can be easily turned on or off as the needs of your lab change for ultimate flexibility in operation and ease-of-use. When enabled, the System Access feature allows assignment of a system administrator and up to 10 user accounts with customizable access to instrument settings, measurement settings, calibrations and logs. Use the password protection to secure selected meter functions, while leaving other functions accessible for all users. The System Access feature allows you to customize meter access to meet your specific security and data reporting needs.