Wash Solution Concentrate (20X), 15 ml
Product Image
Invitrogen™

Wash Solution Concentrate (20X), 15 ml

Wash Solution Concentrate (20X) (contains 0.1% sodium azide) is used to carry out procedures with Luminex™ multiplex panels. Wash SolutionRead more
Have Questions?
Catalog NumberQuantity
WB0415 mL
Catalog number WB04
Price (USD)
33.04
15 ml
In stock
Add to cart
Quantity:
15 mL
Price (USD)
33.04
15 ml
Add to cart
Wash Solution Concentrate (20X) (contains 0.1% sodium azide) is used to carry out procedures with Luminex™ multiplex panels. Wash Solution Concentrate (20X) is designed to meet the additional solution volume often needed when protocols are carried out using an automated system. Please refer to the manual provided with each kit for details on the use of the Wash Solution Concentrate (20X).

Magnetic Beads Let You Do More with Your Time
Luminex™ multiplex panels are built on both polystyrene and magnetic beads and assays for both require the use of Wash Solution Concentrate (20X) as part of their protocols. Magnetic bead-based assays enable automation, help decrease hands-on time, and increase throughput and precision. Using the fast and easy Luminex™ protocols, you can get your results in as little as 3.5 hours.

For research use only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Luminex Instruments, FlexMAP 3D System, MAGPIX System, Luminex 200 System
FormLiquid
Product LineMAGPIX™
Shipping ConditionWet Ice
Product TypeWash Buffer
Quantity15 mL
Unit Size15 ml
Contents & Storage
Wash Solution Concentrate contains:15 mL of 20X wash solution
  • Store at 15°C to 30°C
  • Documents & Downloads

    Certificates

    Safety Data Sheets

    Share catalog number, name or link