Invitrogen™

WesternBreeze™ Chemiluminescent Kit, anti-goat

Catalog number: WB7108
Have Questions?
Invitrogen™

WesternBreeze™ Chemiluminescent Kit, anti-goat

Catalog number: WB7108
Have Questions?
Catalog Number
WB7108
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
WB71081 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
WesternBreeze® Chemiluminescent Kits detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directly from solution (dot blots). Detection is accomplished with a ready-to-use CDP-Star® chemiluminescent substrate for alkaline phosphatase. Protein bands can be captured either by X-ray film or CDP-Star® compatible imaging system. WesternBreeze® Chemiluminescent Kit offers:

• High specificity, clean background
• Ultra-sensitivity-femtogram levels detectable
• Long-lasting signals-up to 5 days
• Results in less than 3 hours
• Permanent photographic image
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity
Goat
Shipping Condition
Wet Ice
Substrate Type
AP (Alkaline Phosphatase) Substrate
Detection Method
Chemiluminescent
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
WesternBreeze™
Product Type
Western Blot Kit
Cross Reactivity
Goat

Contents & Storage

The WesternBreeze™ Chemiluminescent Kits include blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chemiluminescent substrate, wash solutions, incubation trays, pre-cut filter papers, polyester sheet for even substrate development on the membrane. Each kit contains complete reagents for 20 blots. Store the kits at +4°C. Guaranteed stable for 6 months when properly stored.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text