Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Adapters | Includes Adapters |
For Use With (Equipment) | Bio-Rad Criterion Cell |
Gel Percentage | 8% |
Gel Size | Midi |
Gel Thickness | 1.0 mm |
Gel Type | Bis-Tris |
Recommended Applications | Denaturing |
Sample Loading Volume | Up to 15 μL |
Separation Range | 3.5 to 180 kDa |
Separation Type | Denaturing |
Shelf Life | 12 Months |
Wells | 26-well, w/adapters |
Mode of Separation | Molecular Weight |
Product Line | NuPAGE™ |
Quantity | 10 Gels/Box |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 13 cm |
Unit Size | 10 gels (1 box) |
Adapters | No Adapters |
For Use With (Equipment) | XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank |
Gel Percentage | 8% |
Gel Size | Midi |
Gel Thickness | 1.0 mm |
Gel Type | Bis-Tris |
Recommended Applications | Denaturing |
Sample Loading Volume | 45 μL + 15 μL |
Separation Range | 3.5 to 260 kDa |
Separation Type | Denaturing |
Shelf Life | 12 Months |
Wells | 12 + 2-well |
Mode of Separation | Molecular Weight |
Product Line | NuPAGE™ |
Quantity | 10 Gels/Box |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 13 cm |
Unit Size | 10 gels (1 box) |
Adapters | Includes Adapters |
For Use With (Equipment) | Bio-Rad Criterion Cell |
Gel Percentage | 8% |
Gel Size | Midi |
Gel Thickness | 1.0 mm |
Gel Type | Bis-Tris |
Recommended Applications | Denaturing |
Sample Loading Volume | 45 μL + 15 μL |
Separation Range | 3.5 to 260 kDa |
Separation Type | Denaturing |
Shelf Life | 12 Months |
Wells | 12 + 2-well, w/adapters |
Mode of Separation | Molecular Weight |
Product Line | NuPAGE™ |
Quantity | 10 Gels/Box |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 13 cm |
Unit Size | 10 gels (1 box) |
Adapters | No Adapters |
For Use With (Equipment) | XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank |
Gel Percentage | 8% |
Gel Size | Midi |
Gel Thickness | 1.0 mm |
Gel Type | Bis-Tris |
Recommended Applications | Denaturing |
Sample Loading Volume | Up to 25 μL |
Separation Range | 3.5 to 260 kDa |
Separation Type | Denaturing |
Shelf Life | 12 Months |
Wells | 20-well |
Mode of Separation | Molecular Weight |
Product Line | NuPAGE™ |
Quantity | 10 Gels/Box |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 13 cm |
Unit Size | 10 gels (1 box) |
Adapters | Includes Adapters |
For Use With (Equipment) | Bio-Rad Criterion Cell |
Gel Percentage | 8% |
Gel Size | Midi |
Gel Thickness | 1.0 mm |
Gel Type | Bis-Tris |
Recommended Applications | Denaturing |
Sample Loading Volume | Up to 25 μL |
Separation Range | 3.5 to 260 kDa |
Separation Type | Denaturing |
Shelf Life | 12 Months |
Wells | 20-well, w/adapters |
Mode of Separation | Molecular Weight |
Product Line | NuPAGE™ |
Quantity | 10 Gels/Box |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 13 cm |
Unit Size | 10 gels (1 box) |
Adapters | No Adapters |
For Use With (Equipment) | XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank |
Gel Percentage | 8% |
Gel Size | Midi |
Gel Thickness | 1.0 mm |
Gel Type | Bis-Tris |
Recommended Applications | Denaturing |
Sample Loading Volume | Up to 15 μL |
Separation Range | 3.5 to 260 kDa |
Separation Type | Denaturing |
Shelf Life | 12 Months |
Wells | 26-well |
Mode of Separation | Molecular Weight |
Product Line | NuPAGE™ |
Quantity | 10 Gels/Box |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 13 cm |
Unit Size | 10 gels (1 box) |
Catalog Number | Specifications | Unit Size | Wells | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
WG1003A | 10 gels (1 box) | 26-well, w/adapters | 210.65 Online price 223.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
WG1001BOX | 10 gels (1 box) | 12 + 2-well | 199.65 Online price 212.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
WG1001A | 10 gels (1 box) | 12 + 2-well, w/adapters | 211.65 Online price 224.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
WG1002BOX | 10 gels (1 box) | 20-well | 199.65 Online price 212.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
WG1002A | 10 gels (1 box) | 20-well, w/adapters | 211.65 Online price 224.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
WG1003BOX | 10 gels (1 box) | 26-well | 199.65 Online price 212.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features of NuPAGE Bis-Tris gels:
• Better protein integrity—optimized sample preparation process preserves your proteins
• Wide ranges of molecular weight separation—select the right gel and running buffer to get the optimal separation of your proteins
• Faster run times—get separation of your proteins in as little as 35 minutes
• Longer shelf life—NuPAGE Bis-Tris gels can be stored for at least 12 months at room temperature
Choose the right NuPAGE Bis-Tris gel for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. NuPAGE Bis-Tris protein gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and a 4–12% gradient. Gels come in two sizes: mini (8 cm x 8 cm) or midi (8.7 cm x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Bis-Tris gels also come in multiple well formats.
NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Sample Reducing Agent (NP0004). Use NuPAGE Antioxidant (NP0005) in the running buffer to maintain the reduced state of the proteins during the run and to allow maximum band sharpness. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.
NuPAGE Bis-Tris Midi gels can be used with the XCell SureLock Midi Cell gel apparatuses (WR0100) or conveniently with the Bio-Rad Criterion Cell using our adapters (WA0999).
For transfer of proteins to a membrane, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001). Alternatively, traditional wet transfer can be performed using the Bio-Rad Criterion Cell using NuPAGE Transfer buffer (NP00061).