Gibco™

Rat (Wistar) Cryopreserved Hepatocytes, Plateable Male

Catalog number: WICP10
Have Questions?
Gibco™

Rat (Wistar) Cryopreserved Hepatocytes, Plateable Male

Catalog number: WICP10
Have Questions?
Catalog Number
WICP10
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
WICP101 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
These plateable cryopreserved male rat (Wistar) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.

Cells per vial: 4–8 million

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:

• Phase I and II enzyme activities
• Proper morphology
• Viabilities generally >75%
• Attachment efficiency
• Monolayer formation and integrity

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links:

Learn more about our cryopreserved hepatocytes

Learn about our full line of hepatocytes,

Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Hepatocytes
Form
Cryopreserved
Gender
Male
Species
Rat
Donor Source
Pooled
No. of Cells
4 to 8 Million
Product Line
Gibco™
Product Type
Hepatocytes

Contents & Storage

Cryopreserved Hepatocytes stored in liquid nitrogen (LN2).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text