Invitrogen™

Novex™ 4 to 20% Tris-Glycine Plus, 1.0 mm, Midi Protein Gels

Catalog number: WXP42020BOXA
Have Questions?
Invitrogen™

Novex™ 4 to 20% Tris-Glycine Plus, 1.0 mm, Midi Protein Gels

Catalog number: WXP42020BOXA
Have Questions?
Novex Tris-Glycine Plus Midi Protein gels are improved polyacrylamide gels based on traditional Laemmli protein electrophoresis technology, allowing the use of Laemmli sample and running buffers. The gels offer reproducible separation of a wide range of proteins into well-resolved bands. Novex Tris-Glycine Plus Midi gels are a 1.0-mm thick, wider format (8 cm x 13 cm) gel for higher throughput electrophoresis of protein samples.
 
Catalog Number
WXP42020BOXA
Unit Size
10 gels (1 box)
Wells
20-well, w/adapters
Price (USD)
Availability
-
Full specifications
Quantity
AdaptersIncludes Adapters
For Use With (Equipment)Bio-Rad Criterion Cell
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 25 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life6-7 Months
Wells20-well, w/adapters
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
Catalog Number
Unit Size
10 gels (1 box)
Wells
12 + 2-well
Price (USD)
Availability
-
Full specifications
Quantity
AdaptersNo Adapters
For Use With (Equipment)XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 45 μL + 15 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells12 + 2-well
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
Catalog Number
Unit Size
10 gels (1 box)
Wells
12 + 2-well, w/adapters
Price (USD)
Availability
-
Full specifications
Quantity
AdaptersIncludes Adapters
For Use With (Equipment)Bio-Rad Criterion Cell
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 45 μL + 15 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells12 + 2-well, w/adapters
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
Catalog Number
Unit Size
10 gels (1 box)
Wells
20-well
Price (USD)
Availability
-
Full specifications
Quantity
AdaptersNo Adapters
For Use With (Equipment)XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 25 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells20-well
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
Catalog Number
Unit Size
10 gels (1 box)
Wells
26-well
Price (USD)
Availability
-
Full specifications
Quantity
AdaptersNo Adapters
For Use With (Equipment)XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 15 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells26-well
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
Catalog Number
Unit Size
10 gels (1 box)
Wells
26-well, w/adapters
Price (USD)
Availability
-
Full specifications
Quantity
AdaptersIncludes Adapters
For Use With (Equipment)Bio-Rad Criterion Cell
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 15 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells26-well, w/adapters
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeWellsPrice (USD)AvailabilityQuantity
WXP42020BOXAFull specifications
10 gels (1 box)20-well, w/adapters
-
AdaptersIncludes Adapters
For Use With (Equipment)Bio-Rad Criterion Cell
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 25 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life6-7 Months
Wells20-well, w/adapters
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
WXP42012BOXFull specifications
10 gels (1 box)12 + 2-well
-
AdaptersNo Adapters
For Use With (Equipment)XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 45 μL + 15 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells12 + 2-well
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
WXP42012BOXAFull specifications
10 gels (1 box)12 + 2-well, w/adapters
-
AdaptersIncludes Adapters
For Use With (Equipment)Bio-Rad Criterion Cell
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 45 μL + 15 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells12 + 2-well, w/adapters
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
WXP42020BOXFull specifications
10 gels (1 box)20-well
-
AdaptersNo Adapters
For Use With (Equipment)XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 25 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells20-well
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
WXP42026BOXFull specifications
10 gels (1 box)26-well
-
AdaptersNo Adapters
For Use With (Equipment)XCell4 SureLock Midi-Cell, SureLock Tandem Midi Gel Tank
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 15 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells26-well
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
WXP42026BOXAFull specifications
10 gels (1 box)26-well, w/adapters
-
AdaptersIncludes Adapters
For Use With (Equipment)Bio-Rad Criterion Cell
Gel Percentage4 to 20%
Gel SizeMidi
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Sample Loading VolumeUp to 15 μL
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Shelf Life12 Months
Wells26-well, w/adapters
Mode of SeparationMolecular Weight
Product LineNovex™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)13 cm
Unit Size10 gels (1 box)
Showing 6 of 6

Features of Novex Tris-Glycine Plus gels include:
Improved shelf life—store gels for up to 12 months at 4°C
Flexibility—use for native and denaturing protein assays
Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Choose the right gel format for your experiments
Novex Tris-Glycine Plus Midi gels come in fixed concentrations of 10% and 12%, as well as gradient concentrations of 4–12%, 4–20%, and 8–16%. Select from our many well formats, including 12+2-, 20-, and 26-well.

Run your proteins in native or denatured form
Novex Tris-Glycine Plus gels do not contain SDS and so can be used to separate proteins in native or denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26754). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672). The gels can be run using our XCell4 SureLock Midi-Cell (WR0100) or conveniently with the Bio-Rad Criterion Cell using our adapters (WA0999).

For transfer of proteins to a membrane, we recommend using the Novex Tris-Glycine Transfer Buffer (LC3675) when performing traditional wet transfer. Rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

For Research Use Only. Not for use in diagnostic procedures.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text