Novex™ Tris-Glycine Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format
Save now and restock on reagents and precast protein gels! Click here for more information.
Novex™ Tris-Glycine Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format
Invitrogen™

Novex™ Tris-Glycine Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format

Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWellsQuantity
XP00102BOX12-well10 Gels/Box
XP00100PK210-well2 Gels/Box
XP00100BOX10-well10 Gels/Box
XP00105BOX15-well10 Gels/Box
4 Options
Catalog number XP00102BOX
Price (USD)/ 10 gels (1 box)
124.65
Online exclusive
134.00 
Save 9.35 (7%)
-
Add to cart
Wells:
12-well
Quantity:
10 Gels/Box
Price (USD)/ 10 gels (1 box)
124.65
Online exclusive
134.00 
Save 9.35 (7%)
Add to cart
Novex™ Tris-Glycine Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format
Catalog numberXP00102BOX
Price (USD)/ 10 gels (1 box)
124.65
Online exclusive
134.00 
Save 9.35 (7%)
-
Add to cart
Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity. Novex Tris-Glycine Mini Gels are based on traditional Laemmli protein electrophoresis that allow the use of Laemmli sample and running buffers.

Features of Novex Tris-Glycine gels, WedgeWell format, include:
Improved shelf life—store gels for up to 8 months at 4°C
Diversity—use for native and denaturing protein assays
Wedge-shaped wells—easily load up to two times more sample volume
Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Choose the right gel format for your experiments
Novex Tris-Glycine gels, WedgeWell format, come in a variety of fixed concentrations from 6% to 18%, as well as gradients with ranges of 4–12%, 4–20%, 8–16%, and 10–20%. You can select from our many well formats, including 10-, 12-, and 15-well, and different pack sizes.

Run your proteins in native or denatured form
Novex Tris-Glycine gels, WedgeWell format, do not contain SDS and can be used to run your proteins in native or in denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26755). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672).

Convenient to use
Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For transferring your proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) when preforming traditional wet transfer using the Invitrogen Mini Blot Module (B1000). Transfers can also be performed with rapid semi-dry transfer using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage10%
Gel SizeMini
Gel Thickness1.0 mm
Gel TypeTris-Glycine Plus
Recommended ApplicationsDenaturing, Native
Sample Loading VolumeUp to 45 μL
Separation Range30 to 250 kDa
Separation TypeDenaturing, Native
Shelf Life8 Months
Wells12-well
Mode of SeparationMolecular Weight
Product LineNovex™, WedgeWell™
Quantity10 Gels/Box
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Length (Metric)8 cm
Width (Metric)8 cm
Unit Size10 gels (1 box)