Search Thermo Fisher Scientific
Obtain accurate results for antifungal susceptibility of invasive Candida infections to guide therapy with this plate.
Obtain accurate results for antifungal susceptibility of invasive Candida infections to guide therapy with Thermo Scientific™ Sensititre™ YeastOne™ YO4IVD AST Plate with Rezafungin. Report and track echinocandin susceptibility against Candida spp. and make informed decisions to step-down patient therapy with this FDA-cleared panel.
Simplify and consolidate antifungal susceptibility testing on a single plate with Sensititre YeastOne YO4IVD AST Plate with Rezafungin. Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy, and consistent endpoint determination with visual read option for efficient workflow and accurate results.