Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
P2-SC | Spacer Set (2 sides, 1 bottom); 1.0cmW x 0.8mm thickness |
P10-20G | Blank glass; 20cmL x 20cmW x 1/8 in. thickness |
P10-20R | Notched glass; For P2-CST/P10DS; 18cmL x 20cmW x 3/16 in. thickness |
P10-CL | Side Clamps; 1 pair |
P10DS-001 | Upper Buffer Chamber Assembly |
P10DS-006 | Blocking Plate |
P10DS-008 | Lower Buffer Chamber |
P10DS-012 | Super Safe Lid with Attached Power Supply Leads |
P2-SD | Spacer Set (2 sides, 1 bottom); 1.0cmW x 1.5mm thickness |
Keep your dual gel system running with Thermo Scientific™ Owl™ P10DS Dual Gel System Replacement Parts. Parts are for use with the Owl™ P10DS Dual Gel System, an optimal device for a large number of samples that need extended separation and mobility shift assays.