Thermo Scientific Chemicals

Ácido 3,4-diaminobenzoico, 97 %, Thermo Scientific Chemicals

Número de catálogo: 112101000
100 g, Frasco de plástico, Each
Thermo Scientific Chemicals

Ácido 3,4-diaminobenzoico, 97 %, Thermo Scientific Chemicals

Número de catálogo: 112101000
100 g, Frasco de plástico, Each
Cantidad
Envase
Número de catálogo: 112101000
Precio (ARS)

Identificadores químicos

CAS
619-05-6
IUPAC Name
3,4-diaminobenzoate
Molecular Formula
C7H7N2O2
InChI Key
HEMGYNNCNNODNX-UHFFFAOYSA-M
SMILES
NC1=CC=C(C=C1N)C([O-])=O
Appearance (Color)
Brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=96.0 %
Titration with NaOH
96.0 to 104.0 %

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text