Thermo Scientific Chemicals

3,5-Dinitroanilina, 98 %, Thermo Scientific Chemicals

Número de catálogo: 116940100
10 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

3,5-Dinitroanilina, 98 %, Thermo Scientific Chemicals

Número de catálogo: 116940100
10 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 116940100
Precio (USD)

Identificadores químicos

CAS
618-87-1
IUPAC Name
3,5-dinitroaniline
Molecular Formula
C6H5N3O4
InChI Key
MPBZUKLDHPOCLS-UHFFFAOYSA-N
SMILES
NC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Appearance (Color)
Yellow to light brown
Melting point
159°C to 163°C
Appearance (Form)
Powder
HPLC
>=97.5 %
Infrared spectrum
Conforms

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text