4-Fluorofenil sulfona, 99 %, Thermo Scientific Chemicals
4-Fluorofenil sulfona, 99 %, Thermo Scientific Chemicals
4-Fluorofenil sulfona, 99 %, Thermo Scientific Chemicals
4-Fluorofenil sulfona, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorofenil sulfona, 99 %, Thermo Scientific Chemicals

CAS: 383-29-9 | C12H8F2O2S | 254.25 g/mol
Have Questions?
Número de catálogoCantidadEnvase
11962025025 gFrasco de vidrio
Número de catálogo 119620250
Precio (ARS)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS383-29-9
IUPAC Name1-fluoro-4-(4-fluorobenzenesulfonyl)benzene
Molecular FormulaC12H8F2O2S
InChI KeyPLVUIVUKKJTSDM-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Melting point97°C to 100°C
Appearance (Color)White to brown
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Powder or crystals
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0458537Certificate of Analysis18 dic 2023119620250, 119621000
A0455872Certificate of Analysis28 ago 2023119620250, 119621000
A0455336Certificate of Analysis25 ago 2023119620250, 119621000
A0386546Certificate of Analysis12 dic 2022119620250, 119621000
A0446555Certificate of Analysis11 oct 2022119620250, 119621000
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection