Thermo Scientific Chemicals

Bencilcloroformiato, 3 M (50 % en peso) solución en tolueno, Thermo Scientific Chemicals

Número de catálogo: 124495000
500 mL, Frasco de vidrio, Each
Thermo Scientific Chemicals

Bencilcloroformiato, 3 M (50 % en peso) solución en tolueno, Thermo Scientific Chemicals

Número de catálogo: 124495000
500 mL, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 124495000
Precio (USD)

Identificadores químicos

CAS
501-53-1, 108-88-3, 100-44-7
Appearance (Color)
Clear light yellow to light pink
GC
45 to 53 % w/w
Appearance (Form)
Solution

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reactivo de acoplamiento
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text