Thermo Scientific Chemicals

Etil 1-bromociclobutanocarboxilato, 96 %, Thermo Scientific Chemicals

Número de catálogo: 188710050
5 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

Etil 1-bromociclobutanocarboxilato, 96 %, Thermo Scientific Chemicals

Número de catálogo: 188710050
5 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 188710050
Precio (ARS)

Identificadores químicos

CAS
35120-18-4
IUPAC Name
ethyl 1-bromocyclobutane-1-carboxylate
Molecular Formula
C7H11BrO2
InChI Key
UTVNSHXHFRIXMM-UHFFFAOYSA-N
SMILES
CCOC(=O)C1(Br)CCC1
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4700 to 1.4720 (20°C, 589 nm)
GC
>=95 %
Appearance (Form)
Liquid

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text