1-Octeno 99+ %, Thermo Scientific Chemicals
1-Octeno 99+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Octeno 99+ %, Thermo Scientific Chemicals

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 mL
2.5 L
Envase:
Frasco de vidrio
Número de catálogo 301255000
Precio (USD)/ Each
-
Cantidad:
500 mL
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS111-66-0
IUPAC Nameoct-1-ene
Molecular FormulaC8H16
InChI KeyKWKAKUADMBZCLK-UHFFFAOYSA-N
SMILESCCCCCCC=C
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Appearance (Form)Clear liquid
GC>=99.0 %
Water=<0.005 %
Refractive index1.4080 to 1.4100 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only