Thermo Scientific Chemicals

DL-6-Metoxi-α-metil-2-naftalenometanol, 98 %, Thermo Scientific Chemicals

Número de catálogo: 331500250
25 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

DL-6-Metoxi-α-metil-2-naftalenometanol, 98 %, Thermo Scientific Chemicals

Número de catálogo: 331500250
25 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 331500250
Precio (ARS)

Identificadores químicos

CAS
77301-42-9
IUPAC Name
(1R)-1-(6-methoxynaphthalen-2-yl)ethan-1-ol
Molecular Formula
C13H14O2
InChI Key
OUVJWFRUESFCCY-SECBINFHSA-N
SMILES
COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O
Appearance (Color)
White to yellow to light beige
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder
Melting point
111°C to 114°C
Infrared spectrum
Conforms

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text