Thermo Scientific Chemicals

Ácido 5-pirimidinilborónico, 97 %, puede contener cantidades variables de anhídrido, Thermo Scientific Chemicals

Número de catálogo: 451700250
25 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

Ácido 5-pirimidinilborónico, 97 %, puede contener cantidades variables de anhídrido, Thermo Scientific Chemicals

Número de catálogo: 451700250
25 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 451700250
Precio (USD)

Identificadores químicos

CAS
109299-78-7
IUPAC Name
(pyrimidin-5-yl)boronic acid
Molecular Formula
C4H5BN2O2
InChI Key
HZFPPBMKGYINDF-UHFFFAOYSA-N
SMILES
OB(O)C1=CN=CN=C1
Appearance (Color)
White to off-white
HPLC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Form)
Solid

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text