Thermo Scientific Chemicals

Cloruro de 3-bromobenzoílo, 98 %, Thermo Scientific Chemicals

Número de catálogo: A15256.30
250 g, Each
Thermo Scientific Chemicals

Cloruro de 3-bromobenzoílo, 98 %, Thermo Scientific Chemicals

Número de catálogo: A15256.30
250 g, Each
Cantidad
Número de catálogo: A15256.30
también denominado A15256-30
Precio (USD)

Identificadores químicos

CAS
1711-09-7
IUPAC Name
3-bromobenzoyl chloride
Molecular Formula
C7H4BrClO
InChI Key
PBOOZQFGWNZNQE-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=CC(Br)=C1
Form
Liquid
Refractive Index
1.5935-1.5975 @ 20°C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to yellow

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text