Métoxido de germanio(IV), 98 %, Thermo Scientific Chemicals
Métoxido de germanio(IV), 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Métoxido de germanio(IV), 98 %, Thermo Scientific Chemicals

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Número de catálogo A18437.06
también denominado A18437-06
Precio (USD)/ Each
-
Cantidad:
5 g
Pedido a granel o personalizado
Identificadores químicos
CAS992-91-6
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear, colorless
FormLiquid
Assay (unspecified)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only