Thermo Scientific Chemicals

3-Cloro-2,6-dimetilanilina, 99 %, Thermo Scientific Chemicals

Número de catálogo: B22232.18
50 g, Each
Thermo Scientific Chemicals

3-Cloro-2,6-dimetilanilina, 99 %, Thermo Scientific Chemicals

Número de catálogo: B22232.18
50 g, Each
Cantidad
Número de catálogo: B22232.18
también denominado B22232-18
Precio (USD)

Identificadores químicos

CAS
26829-77-6
IUPAC Name
3-chloro-2,6-dimethylaniline
Molecular Formula
C8H10ClN
InChI Key
FGMXFOTYCHZCLA-UHFFFAOYSA-N
SMILES
CC1=CC=C(Cl)C(C)=C1N
Appearance (Color)
White to dark brown to pale brown or pink
Form
Fused solid or clear liquid as melt
Assay (GC)
≥98.5%
Melting Point (clear melt)
28.0-35.0?C

Descripción

3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text