Ácido 2-fluoro-alfa-metil-4-bifenilacético, 99 %, Thermo Scientific Chemicals
Ácido 2-fluoro-alfa-metil-4-bifenilacético, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido 2-fluoro-alfa-metil-4-bifenilacético, 99 %, Thermo Scientific Chemicals

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
1 g
Número de catálogo B22603.14
también denominado B22603-14
Precio (USD)/ Each
-
Cantidad:
25 g
Pedido a granel o personalizado
Identificadores químicos
CAS5104-49-4
IUPAC Name2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
Molecular FormulaC15H13FO2
InChI KeySYTBZMRGLBWNTM-UHFFFAOYNA-N
SMILESCC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Melting Point (clear melt)111.5-115.5?C
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
A cyclooxygenase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only