Thermo Scientific Chemicals

2,4-Difluoro-3-metoxianilina, 97 %, Thermo Scientific Chemicals

Número de catálogo: H26489.03
1 g, Each
Thermo Scientific Chemicals

2,4-Difluoro-3-metoxianilina, 97 %, Thermo Scientific Chemicals

Número de catálogo: H26489.03
1 g, Each
Cantidad
Número de catálogo: H26489.03
también denominado H26489-03
Precio (ARS)
Have Questions?

Identificadores químicos

CAS
886499-08-7
Appearance (Color)
Clear colorless to yellow to brown
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5105-1.5155 @ 20°C

Descripción

2,4-Difluoro-3-methoxyaniline is a fluorinated compound used as a building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
La 2,4-difluoro-3-metoxianilina es un compuesto fluorado utilizado como compuesto básico en la síntesis orgánica.

Solubilidad
No miscible con agua.

Notas
Incompatible con los agentes oxidantes fuertes.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text