Thermo Scientific Chemicals

Ácido trans-4-(Boc-amino)ciclohexanoacético, 97 %, Thermo Scientific Chemicals

Número de catálogo: H62058.03
1 g, Each
Thermo Scientific Chemicals

Ácido trans-4-(Boc-amino)ciclohexanoacético, 97 %, Thermo Scientific Chemicals

Número de catálogo: H62058.03
1 g, Each
Cantidad
Número de catálogo: H62058.03
también denominado H62058-03
Precio (USD)

Descripción

trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water (1.1 g/L) (25°C)

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text