Fosfato, solución de tampón de 0,5 M, pH 7,4
Fosfato, solución de tampón de 0,5 M, pH 7,4
Thermo Scientific Chemicals

Fosfato, solución de tampón de 0,5 M, pH 7,4

Buffer solution commonly used in biological research. A high buffering capacity reagent used in molecular biology, biochemistry, and chromatography.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500mL
250mL
Número de catálogo J60785.AP
también denominado J60785-AP
Precio (USD)/ Each
-
Cantidad:
500mL
Pedido a granel o personalizado
Especificaciones
ConcentraciónSolución de tampón de 0,5 m
Peso molecular119,976
Cantidad500mL
EnvaseFrasco de plástico
CAS7558-80-7
Ver más
Phosphate-buffered saline (abbreviated PBS) is a buffer solution commonly used in biological research. A high buffering capacity reagent used in molecular biology, biochemistry, and chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contenido y almacenamiento
Temperaturas ambientes