4,4'-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
La 4,4′-dibromobenzofenona se utiliza en la síntesis de derivados del poli(p-fenileno) de alto peso molecular a través de una reacción de policondensación catalizada por paladio.
Solubilidad
Soluble en etanol, 2-propanol, acetona y tolueno. Miscible con agua.
Notas
Sensible a la luz. Incompatible con agentes oxidantes fuertes y ácidos fuertes.
RUO – Research Use Only
General References:
- Mukherjee, S.; Thilagar, P. Recent advances in purely organic phosphorescent materials. Chem. Commun. 2015, 51 (55), 10988-11003.
- Ravoet, J.; Reybroeck, W.; De Graaf, D. C. Pesticides for Apicultural and/or Agricultural Application Found in Belgian Honey Bee Wax Combs. B. Environ. Contam. Tox. 2015, 94 (5), 543-548.