Thermo Scientific Chemicals

Pipecolinato de etilo, + 98 %, Thermo Scientific Chemicals

Número de catálogo: L10532.06
5 g, Each
Thermo Scientific Chemicals

Pipecolinato de etilo, + 98 %, Thermo Scientific Chemicals

Número de catálogo: L10532.06
5 g, Each
Cantidad
Número de catálogo: L10532.06
también denominado L10532-06
Precio (USD)

Identificadores químicos

CAS
15862-72-3
IUPAC Name
(2S)-2-(ethoxycarbonyl)piperidin-1-ium
Molecular Formula
C8H16NO2
InChI Key
SZIKRGHFZTYTIT-ZETCQYMHSA-O
SMILES
CCOC(=O)[C@@H]1CCCC[NH2+]1
Form
Liquid
Refractive Index
1.4540-1.4580 @ 20?C
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to yellow

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text