BactiDisks™ Pseudomonas aeruginosa ATCC™ 10145™
BactiDisks™ Pseudomonas aeruginosa ATCC™ 10145™
Thermo Scientific™
BactiDisks™ Pseudomonas aeruginosa ATCC™ 10145™
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
Have Questions?
Número de catálogoCantidad
R1912610 Disk/Vial
Número de catálogo R19126
Precio (ARS)
-
Solicite un presupuesto
Cantidad:
10 Disk/Vial
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Especificaciones
ATCC®10145™*
DescripciónPseudomonas aeruginosa ATCC™ 10145™
FormatoBactiDisks
Tipo de productoBactidisk
Clase de organismo dianaGram-negative Rods
Tipo de pruebaQualitative QC
Cantidad10 Disk/Vial
Unit SizeEach
Documentos y descargas
Certificados
Hojas de datos de seguridad
Compartir número de catálogo, nombre o enlace.