Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
With its unique, flexible detection scheme, the Thermo Scientific™ Scios™ DualBeam™ offers simultaneous detection of all information with excellent contrast at high acquisition speeds and high resolution at low kV. This makes it ideal for imaging a wide range of samples, including very sensitive materials. Fully automated column alignments limit the need for highly trained operators, while predefined use cases for common conditions give instant productivity to everyone, which is especially relevant to multi-user facilities.
Advanced detection technology is at the very core of the Scios DualBeam. In-lens Thermo Scientific™ Trinity™ detection technology collects all signals simultaneously, saving time and offering distinctly different contrasts to capture the maximum amount of data. An innovative, under-the-lens concentric backscatter detector enhances efficiency, enabling you to select a signal based on its angular distribution to easily separate materials and topographic contrast - even at 20 eV landing energy.
With innovative features designed to increase throughput, precision, and ease of use, our DualBeam is ideal for advanced research and analysis across academic, government, and industrial research environments. With unique User Guidance and step-by-step workflows for common DualBeam applications, the Scios DualBeam provides instant productivity for operators of all experience levels.
Efficient workflows enable the Scios DualBeam to produce accurate results in the shortest possible time. The Amira SDB wizard streamlines the steps necessary for accurate analysis and visualization of 3D data sets acquired on the Scios DualBeam. Correlative Light and Electron Microscopy (CLEM) targeted AutoSlice & View provides a true workflow solution for finding the precise location of the region of interest in a large volume, enabling high-resolution imaging and simplified, high-precision, cryo-TEM sample prep. Such precision offers the highest yield without contamination, even when working with challenging samples.