| SH31217.43 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1 in. ID Silicone, 1 1/2 Tri-Clamps | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 15 ft., 1 in. ID Silicone, 1 1/2 Tri-Clamps | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | Non-aseptic | Unit Size | 1 each |
|
| SH31387.01 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamps | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamps | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | 1 1/2 Tri-Clamps | Material del tubo | Silicone | Unit Size | 1 each |
|
| SH31217.41 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID Silicone, 1 1/2 Tri-Clamps | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID Silicone, 1 1/2 Tri-Clamps | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | Non-aseptic | Unit Size | 1 each |
|
| SH31387.02 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamps | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamps | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | 1 1/2 Tri-Clamps | Material del tubo | Silicone | Unit Size | 1 each |
|
| SH31217.42 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1 in. ID Silicone, 1 1/2 Tri-Clamps | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 10 ft., 1 in. ID Silicone, 1 1/2 Tri-Clamps | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | Non-aseptic | Unit Size | 1 each |
|
| SH31387.03 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamps | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamps | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | 1 1/2 Tri-Clamps | Material del tubo | Silicone | Unit Size | 1 each |
|
| SH31377.01 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Silicone, MPC Insert and MPC Body | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID Silicone, MPC Insert and MPC Body | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | MPC Insert and MPC Body | Material del tubo | Silicone | Unit Size | 1 each |
|
| SH31386.01 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | Non-aseptic | Unit Size | 1 each |
|
| SH31377.02 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID Silicone, MPC Insert and MPC Body | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID Silicone, MPC Insert and MPC Body | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | MPC Insert and MPC Body | Material del tubo | Silicone | Unit Size | 1 each |
|
| SH31386.02 | Especificaciones completas | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | Solicite un presupuesto |
Descripción | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Silicone, 1 1/2 Tri-Clamp and 3/4 Tri-Clamp | Para utilizar con (aplicación) | Monoclonal antibody production, upstream and downstream applications | Cantidad | Each | Tipos de conector | Non-aseptic | Unit Size | 1 each |
|